Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8123782-8124055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4A
Entrez Gene ID
50856
Gene Name
C-type lectin domain family 4 member A
Gene Aliases
CD367, CLECSF6, DCIR, DDB27, HDCGC13P, LLIR
UniGene
Hs.504657
Gene Chromosome Location
Chr.12: 8102932-8138607 on Build GRCh38
Transcripts
NM_016184.3 NM_194447.2 NM_194448.2 NM_194450.2 XM_017019382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1993783 c.8C>T p.S3L Chr.12: 8123886-8123886 on Build GRCh38
6871326 c.54C>T p.S18S Chr.12: 8123932-8123932 on Build GRCh38
376016 c.81A>T p.A27A Chr.12: 8123959-8123959 on Build GRCh38
1364552 c.9G>A p.S3S Chr.12: 8123887-8123887 on Build GRCh38
6138416 c.65A>G p.N22S Chr.12: 8123943-8123943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750821923 Chr.12: 8123880-8123880 on Build GRCh38
rs753851877 Chr.12: 8123949-8123949 on Build GRCh38
rs766924798 Chr.12: 8123893-8123893 on Build GRCh38
rs771647674 Chr.12: 8123868-8123868 on Build GRCh38
rs775187479 Chr.12: 8123875-8123875 on Build GRCh38
rs768136096 Chr.12: 8123985-8123985 on Build GRCh38
rs773649897 Chr.12: 8123886-8123886 on Build GRCh38
rs764122873 Chr.12: 8123981-8123981 on Build GRCh38
rs36113649 Chr.12: 8123943-8123943 on Build GRCh38
rs1008183322 Chr.12: 8123817-8123817 on Build GRCh38
rs142534143 Chr.12: 8123940-8123940 on Build GRCh38
rs746541863 Chr.12: 8123879-8123879 on Build GRCh38
rs758182604 Chr.12: 8123979-8123979 on Build GRCh38
rs778687385 Chr.12: 8123861-8123861 on Build GRCh38
rs767679694 Chr.12: 8123919-8123919 on Build GRCh38
rs747619528 Chr.12: 8124004-8124004 on Build GRCh38
rs770505223 Chr.12: 8123938-8123939 on Build GRCh38
rs778281838 Chr.12: 8123969-8123969 on Build GRCh38
rs774885996 Chr.12: 8123899-8123899 on Build GRCh38
rs752929459 Chr.12: 8123939-8123939 on Build GRCh38
rs943991733 Chr.12: 8123870-8123870 on Build GRCh38
rs969829813 Chr.12: 8124023-8124023 on Build GRCh38
rs776283379 Chr.12: 8123996-8123996 on Build GRCh38
rs915605906 Chr.12: 8124021-8124021 on Build GRCh38
rs745624340 Chr.12: 8123867-8123867 on Build GRCh38
rs756596934 Chr.12: 8123894-8123894 on Build GRCh38
rs367759558 Chr.12: 8123983-8123983 on Build GRCh38
rs968972991 Chr.12: 8123878-8123878 on Build GRCh38
rs757132340 Chr.12: 8123856-8123856 on Build GRCh38
rs7295783 Chr.12: 8123836-8123836 on Build GRCh38
rs774082428 Chr.12: 8124012-8124012 on Build GRCh38
rs758914176 Chr.12: 8123958-8123958 on Build GRCh38
rs184359922 Chr.12: 8123887-8123887 on Build GRCh38
rs79958734 Chr.12: 8123945-8123945 on Build GRCh38
rs759928998 Chr.12: 8123904-8123904 on Build GRCh38
rs745714090 Chr.12: 8123963-8123963 on Build GRCh38

View Full Product Details