Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 30900169-30900440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCDC5
Entrez Gene ID
100506627
Gene Name
doublecortin domain containing 5
Gene Aliases
-
UniGene
Hs.530438
Gene Chromosome Location
Chr.11: 30863603-30992686 on Build GRCh38
Transcripts
NM_020869.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5143326 c.714A>T p.L238L Chr.11: 30900356-30900356 on Build GRCh38
4532450 c.634G>A p.D212N Chr.11: 30900436-30900436 on Build GRCh38
466721 c.724+2T>C p.? Chr.11: 30900344-30900344 on Build GRCh38
4532449 c.1801G>A p.D601N Chr.11: 30900436-30900436 on Build GRCh38
4977162 c.1844C>G p.P615R Chr.11: 30900393-30900393 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772288149 Chr.11: 30900358-30900358 on Build GRCh38
rs771487302 Chr.11: 30900309-30900309 on Build GRCh38
rs556194727 Chr.11: 30900334-30900334 on Build GRCh38
rs16921560 Chr.11: 30900218-30900218 on Build GRCh38
rs751300083 Chr.11: 30900405-30900405 on Build GRCh38
rs1036189647 Chr.11: 30900275-30900275 on Build GRCh38
rs750424489 Chr.11: 30900328-30900328 on Build GRCh38
rs763223087 Chr.11: 30900316-30900316 on Build GRCh38
rs145957720 Chr.11: 30900371-30900371 on Build GRCh38
rs201129626 Chr.11: 30900391-30900391 on Build GRCh38
rs139526404 Chr.11: 30900266-30900266 on Build GRCh38
rs758787077 Chr.11: 30900334-30900335 on Build GRCh38
rs975656903 Chr.11: 30900208-30900208 on Build GRCh38
rs947469892 Chr.11: 30900274-30900274 on Build GRCh38
rs186492871 Chr.11: 30900228-30900228 on Build GRCh38
rs547968575 Chr.11: 30900271-30900271 on Build GRCh38
rs968323565 Chr.11: 30900199-30900199 on Build GRCh38
rs555812486 Chr.11: 30900243-30900243 on Build GRCh38
rs773192784 Chr.11: 30900379-30900379 on Build GRCh38
rs896060197 Chr.11: 30900401-30900401 on Build GRCh38
rs760849425 Chr.11: 30900369-30900369 on Build GRCh38
rs752860736 Chr.11: 30900278-30900278 on Build GRCh38
rs775462661 Chr.11: 30900367-30900367 on Build GRCh38
rs35235694 Chr.11: 30900303-30900304 on Build GRCh38
rs1044949207 Chr.11: 30900324-30900324 on Build GRCh38
rs753393262 Chr.11: 30900320-30900320 on Build GRCh38
rs112921806 Chr.11: 30900315-30900315 on Build GRCh38
rs779351420 Chr.11: 30900310-30900310 on Build GRCh38
rs899003380 Chr.11: 30900295-30900295 on Build GRCh38
rs1043463299 Chr.11: 30900302-30900302 on Build GRCh38

View Full Product Details