Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1844809-1845078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEIOB
Entrez Gene ID
254528
Gene Name
meiosis specific with OB domains
Gene Aliases
C16orf73, gs129
UniGene
Hs.729604
Gene Chromosome Location
Chr.16: 1833983-1872178 on Build GRCh38
Transcripts
NM_001163560.2 NM_152764.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5706025 c.802C>A p.L268M Chr.16: 1844940-1844940 on Build GRCh38
5706026 c.70C>A p.L24M Chr.16: 1844940-1844940 on Build GRCh38
6390419 c.791C>G p.A264G Chr.16: 1844951-1844951 on Build GRCh38
1376213 c.82C>T p.R28* Chr.16: 1844928-1844928 on Build GRCh38
6381570 c.820A>G p.N274D Chr.16: 1844922-1844922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756535144 Chr.16: 1844843-1844843 on Build GRCh38
rs373035025 Chr.16: 1844951-1844951 on Build GRCh38
rs187275503 Chr.16: 1844997-1844997 on Build GRCh38
rs913296809 Chr.16: 1844893-1844893 on Build GRCh38
rs769285815 Chr.16: 1844895-1844895 on Build GRCh38
rs1046615342 Chr.16: 1844937-1844937 on Build GRCh38
rs775376213 Chr.16: 1845007-1845017 on Build GRCh38
rs777521693 Chr.16: 1844968-1844968 on Build GRCh38
rs746739940 Chr.16: 1844880-1844880 on Build GRCh38
rs998367976 Chr.16: 1845011-1845011 on Build GRCh38
rs149488366 Chr.16: 1844958-1844958 on Build GRCh38
rs191995022 Chr.16: 1845007-1845007 on Build GRCh38
rs386787761 Chr.16: 1845029-1845030 on Build GRCh38
rs185686718 Chr.16: 1845028-1845028 on Build GRCh38
rs767852650 Chr.16: 1844833-1844833 on Build GRCh38
rs758460084 Chr.16: 1844868-1844868 on Build GRCh38
rs4644862 Chr.16: 1845030-1845030 on Build GRCh38
rs957929594 Chr.16: 1845018-1845018 on Build GRCh38
rs750799555 Chr.16: 1844836-1844836 on Build GRCh38
rs781104152 Chr.16: 1844995-1844995 on Build GRCh38
rs9806825 Chr.16: 1844851-1844851 on Build GRCh38
rs4598914 Chr.16: 1845029-1845029 on Build GRCh38
rs116866098 Chr.16: 1844990-1844990 on Build GRCh38
rs771558087 Chr.16: 1845002-1845004 on Build GRCh38
rs375920721 Chr.16: 1844885-1844885 on Build GRCh38
rs767975202 Chr.16: 1844913-1844913 on Build GRCh38
rs374328289 Chr.16: 1844994-1844994 on Build GRCh38
rs748087466 Chr.16: 1844863-1844863 on Build GRCh38
rs755310473 Chr.16: 1844947-1844947 on Build GRCh38
rs9806945 Chr.16: 1844960-1844960 on Build GRCh38
rs776088154 Chr.16: 1844889-1844889 on Build GRCh38
rs564132692 Chr.16: 1844927-1844927 on Build GRCh38
rs758581396 Chr.16: 1844964-1844964 on Build GRCh38
rs762369344 Chr.16: 1844828-1844828 on Build GRCh38
rs1030210626 Chr.16: 1845021-1845021 on Build GRCh38
rs775035952 Chr.16: 1844909-1844909 on Build GRCh38
rs773793662 Chr.16: 1844916-1844916 on Build GRCh38
rs555327933 Chr.16: 1844852-1844852 on Build GRCh38
rs9806826 Chr.16: 1844911-1844911 on Build GRCh38
rs779201835 Chr.16: 1844861-1844861 on Build GRCh38
rs745535215 Chr.16: 1844891-1844891 on Build GRCh38
rs58285301 Chr.16: 1845031-1845031 on Build GRCh38
rs745529366 Chr.16: 1844913-1844916 on Build GRCh38
rs575464897 Chr.16: 1844874-1844874 on Build GRCh38

View Full Product Details