Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 81078334-81078559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSS3
Entrez Gene ID
79611
Gene Name
acyl-CoA synthetase short-chain family member 3
Gene Aliases
-
UniGene
Hs.259559
Gene Chromosome Location
Chr.12: 81078018-81257795 on Build GRCh38
Transcripts
NM_024560.2 XM_005269150.2 XM_005269151.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3872588 c.222G>A p.E74E Chr.12: 81078342-81078342 on Build GRCh38
1580421 c.214G>A p.D72N Chr.12: 81078334-81078334 on Build GRCh38
5696922 c.264G>T p.K88N Chr.12: 81078384-81078384 on Build GRCh38
6194633 c.233G>T p.G78V Chr.12: 81078353-81078353 on Build GRCh38
6592827 c.276A>C p.K92N Chr.12: 81078396-81078396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555649704 Chr.12: 81078381-81078381 on Build GRCh38
rs866040700 Chr.12: 81078385-81078385 on Build GRCh38
rs990770199 Chr.12: 81078361-81078361 on Build GRCh38
rs900768470 Chr.12: 81078493-81078493 on Build GRCh38
rs540422865 Chr.12: 81078399-81078399 on Build GRCh38
rs767130847 Chr.12: 81078440-81078440 on Build GRCh38
rs950896312 Chr.12: 81078521-81078521 on Build GRCh38
rs995070078 Chr.12: 81078503-81078503 on Build GRCh38
rs759138967 Chr.12: 81078437-81078437 on Build GRCh38
rs769687967 Chr.12: 81078418-81078418 on Build GRCh38
rs753041243 Chr.12: 81078444-81078444 on Build GRCh38
rs773188180 Chr.12: 81078419-81078419 on Build GRCh38
rs918673276 Chr.12: 81078486-81078486 on Build GRCh38
rs1013913366 Chr.12: 81078353-81078353 on Build GRCh38
rs1028267080 Chr.12: 81078514-81078514 on Build GRCh38
rs141909100 Chr.12: 81078420-81078420 on Build GRCh38
rs779285826 Chr.12: 81078369-81078369 on Build GRCh38
rs749390394 Chr.12: 81078364-81078364 on Build GRCh38
rs766320623 Chr.12: 81078423-81078424 on Build GRCh38
rs189529003 Chr.12: 81078456-81078456 on Build GRCh38
rs935097849 Chr.12: 81078436-81078436 on Build GRCh38
rs1006519890 Chr.12: 81078490-81078490 on Build GRCh38
rs373217798 Chr.12: 81078389-81078389 on Build GRCh38
rs776341687 Chr.12: 81078400-81078400 on Build GRCh38
rs1017856318 Chr.12: 81078492-81078492 on Build GRCh38
rs377599766 Chr.12: 81078422-81078422 on Build GRCh38
rs747945335 Chr.12: 81078416-81078416 on Build GRCh38
rs779089611 Chr.12: 81078474-81078474 on Build GRCh38
rs778310119 Chr.12: 81078354-81078354 on Build GRCh38
rs554106519 Chr.12: 81078351-81078351 on Build GRCh38
rs761091790 Chr.12: 81078448-81078448 on Build GRCh38
rs77681927 Chr.12: 81078459-81078459 on Build GRCh38
rs764563793 Chr.12: 81078451-81078451 on Build GRCh38
rs868824020 Chr.12: 81078384-81078384 on Build GRCh38
rs774022259 Chr.12: 81078434-81078434 on Build GRCh38
rs754308903 Chr.12: 81078453-81078453 on Build GRCh38
rs142563750 Chr.12: 81078484-81078484 on Build GRCh38
rs542100679 Chr.12: 81078476-81078476 on Build GRCh38
rs746050655 Chr.12: 81078382-81078382 on Build GRCh38

View Full Product Details