Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26443032-26443273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPHD2
Entrez Gene ID
57168
Gene Name
aspartate beta-hydroxylase domain containing 2
Gene Aliases
-
UniGene
Hs.567547
Gene Chromosome Location
Chr.22: 26429314-26445012 on Build GRCh38
Transcripts
NM_020437.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1032961 c.985G>A p.A329T Chr.22: 26443159-26443159 on Build GRCh38
1724457 c.1001-9delC p.? Chr.22: 26443088-26443088 on Build GRCh38
4103173 c.1101G>A p.P367P Chr.22: 26443197-26443197 on Build GRCh38
4103171 c.1022G>A p.R341Q Chr.22: 26443118-26443118 on Build GRCh38
4298164 c.1069G>T p.E357* Chr.22: 26443165-26443165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900341210 Chr.22: 26443169-26443169 on Build GRCh38
rs142093449 Chr.22: 26443155-26443155 on Build GRCh38
rs556033981 Chr.22: 26443153-26443153 on Build GRCh38
rs772592077 Chr.22: 26443198-26443198 on Build GRCh38
rs189222529 Chr.22: 26443089-26443089 on Build GRCh38
rs751325098 Chr.22: 26443161-26443161 on Build GRCh38
rs888565479 Chr.22: 26443246-26443246 on Build GRCh38
rs567864544 Chr.22: 26443171-26443171 on Build GRCh38
rs369235514 Chr.22: 26443129-26443129 on Build GRCh38
rs568117916 Chr.22: 26443088-26443088 on Build GRCh38
rs141465218 Chr.22: 26443105-26443105 on Build GRCh38
rs138986249 Chr.22: 26443118-26443118 on Build GRCh38
rs1023027449 Chr.22: 26443201-26443201 on Build GRCh38
rs764905985 Chr.22: 26443152-26443152 on Build GRCh38
rs377666159 Chr.22: 26443197-26443197 on Build GRCh38
rs142309739 Chr.22: 26443092-26443092 on Build GRCh38
rs868194157 Chr.22: 26443196-26443196 on Build GRCh38
rs755260162 Chr.22: 26443087-26443088 on Build GRCh38
rs749581355 Chr.22: 26443173-26443173 on Build GRCh38
rs544809372 Chr.22: 26443202-26443202 on Build GRCh38
rs759301055 Chr.22: 26443231-26443231 on Build GRCh38
rs547974941 Chr.22: 26443168-26443168 on Build GRCh38
rs145158799 Chr.22: 26443117-26443117 on Build GRCh38
rs764629413 Chr.22: 26443232-26443232 on Build GRCh38
rs769413595 Chr.22: 26443084-26443084 on Build GRCh38
rs763956357 Chr.22: 26443156-26443156 on Build GRCh38
rs751508328 Chr.22: 26443250-26443250 on Build GRCh38
rs779101531 Chr.22: 26443188-26443188 on Build GRCh38
rs760429140 Chr.22: 26443065-26443065 on Build GRCh38
rs748242834 Chr.22: 26443218-26443218 on Build GRCh38
rs753942230 Chr.22: 26443088-26443088 on Build GRCh38
rs374053512 Chr.22: 26443177-26443177 on Build GRCh38
rs753359088 Chr.22: 26443086-26443086 on Build GRCh38
rs145940727 Chr.22: 26443115-26443115 on Build GRCh38
rs372650643 Chr.22: 26443253-26443253 on Build GRCh38
rs775015859 Chr.22: 26443238-26443238 on Build GRCh38
rs766153022 Chr.22: 26443071-26443071 on Build GRCh38
rs776417011 Chr.22: 26443229-26443229 on Build GRCh38
rs140180423 Chr.22: 26443221-26443221 on Build GRCh38
rs180855742 Chr.22: 26443137-26443137 on Build GRCh38
rs771483098 Chr.22: 26443228-26443228 on Build GRCh38
rs748733963 Chr.22: 26443103-26443103 on Build GRCh38
rs752737178 Chr.22: 26443090-26443090 on Build GRCh38
rs774719903 Chr.22: 26443101-26443101 on Build GRCh38
rs768733964 Chr.22: 26443094-26443094 on Build GRCh38
rs35429415 Chr.22: 26443247-26443247 on Build GRCh38
rs577698732 Chr.22: 26443242-26443242 on Build GRCh38
rs75079091 Chr.22: 26443093-26443093 on Build GRCh38
rs757048347 Chr.22: 26443164-26443164 on Build GRCh38
rs778916081 Chr.22: 26443112-26443113 on Build GRCh38

View Full Product Details