Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28880528-28880787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLOT2
Entrez Gene ID
2319
Gene Name
flotillin 2
Gene Aliases
ECS-1, ECS1, ESA, ESA1, M17S1
UniGene
Hs.514038
Gene Chromosome Location
Chr.17: 28879339-28897728 on Build GRCh38
Transcripts
NM_004475.2 XM_005257950.3 XM_005257953.2 XM_017024394.1 XM_017024395.1 XM_017024396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7078645 c.1227C>T p.L409L Chr.17: 28880734-28880734 on Build GRCh38
7087550 c.1249-1G>A p.? Chr.17: 28880600-28880600 on Build GRCh38
3969927 c.1223C>T p.A408V Chr.17: 28880738-28880738 on Build GRCh38
7232484 c.1206G>A p.L402L Chr.17: 28880755-28880755 on Build GRCh38
3515518 c.1188C>T p.N396N Chr.17: 28880773-28880773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764712313 Chr.17: 28880703-28880704 on Build GRCh38
rs371925989 Chr.17: 28880667-28880667 on Build GRCh38
rs768005474 Chr.17: 28880707-28880707 on Build GRCh38
rs753921937 Chr.17: 28880641-28880641 on Build GRCh38
rs769470236 Chr.17: 28880691-28880691 on Build GRCh38
rs925839126 Chr.17: 28880745-28880745 on Build GRCh38
rs1029305667 Chr.17: 28880710-28880710 on Build GRCh38
rs775052595 Chr.17: 28880559-28880559 on Build GRCh38
rs761836414 Chr.17: 28880721-28880721 on Build GRCh38
rs201936801 Chr.17: 28880727-28880727 on Build GRCh38
rs1043274063 Chr.17: 28880736-28880736 on Build GRCh38
rs968020529 Chr.17: 28880719-28880719 on Build GRCh38
rs60578192 Chr.17: 28880692-28880692 on Build GRCh38
rs372414269 Chr.17: 28880565-28880565 on Build GRCh38
rs369330831 Chr.17: 28880702-28880702 on Build GRCh38
rs761055985 Chr.17: 28880702-28880703 on Build GRCh38
rs761378408 Chr.17: 28880671-28880671 on Build GRCh38
rs754349002 Chr.17: 28880707-28880710 on Build GRCh38
rs375396199 Chr.17: 28880635-28880635 on Build GRCh38
rs951023348 Chr.17: 28880620-28880620 on Build GRCh38
rs547066451 Chr.17: 28880668-28880668 on Build GRCh38
rs769292693 Chr.17: 28880705-28880705 on Build GRCh38
rs767351713 Chr.17: 28880588-28880588 on Build GRCh38
rs760426450 Chr.17: 28880749-28880749 on Build GRCh38
rs1027859196 Chr.17: 28880602-28880602 on Build GRCh38
rs199581006 Chr.17: 28880594-28880594 on Build GRCh38
rs761867326 Chr.17: 28880581-28880581 on Build GRCh38
rs1060244 Chr.17: 28880553-28880553 on Build GRCh38
rs769399702 Chr.17: 28880556-28880556 on Build GRCh38
rs778108827 Chr.17: 28880687-28880687 on Build GRCh38
rs1027354481 Chr.17: 28880666-28880666 on Build GRCh38
rs376264541 Chr.17: 28880729-28880729 on Build GRCh38
rs906155366 Chr.17: 28880578-28880578 on Build GRCh38
rs748595512 Chr.17: 28880670-28880670 on Build GRCh38
rs77888293 Chr.17: 28880595-28880595 on Build GRCh38
rs776067761 Chr.17: 28880693-28880700 on Build GRCh38
rs751505577 Chr.17: 28880574-28880574 on Build GRCh38
rs774000334 Chr.17: 28880712-28880712 on Build GRCh38
rs773581646 Chr.17: 28880550-28880550 on Build GRCh38
rs946860498 Chr.17: 28880722-28880722 on Build GRCh38

View Full Product Details