Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68022038-68022263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX28
Entrez Gene ID
55794
Gene Name
DEAD-box helicase 28
Gene Aliases
MDDX28
UniGene
Hs.458313
Gene Chromosome Location
Chr.16: 68021274-68023867 on Build GRCh38
Transcripts
NM_018380.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
268996 c.1019T>C p.L340S Chr.16: 68022184-68022184 on Build GRCh38
4062067 c.1136C>T p.A379V Chr.16: 68022067-68022067 on Build GRCh38
1203242 c.1077C>A p.H359Q Chr.16: 68022126-68022126 on Build GRCh38
4890828 c.1003G>A p.E335K Chr.16: 68022200-68022200 on Build GRCh38
6641697 c.1163G>A p.R388H Chr.16: 68022040-68022040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769910674 Chr.16: 68022060-68022060 on Build GRCh38
rs765573829 Chr.16: 68022061-68022062 on Build GRCh38
rs1052882338 Chr.16: 68022057-68022057 on Build GRCh38
rs768369352 Chr.16: 68022071-68022071 on Build GRCh38
rs758615165 Chr.16: 68022152-68022152 on Build GRCh38
rs749263499 Chr.16: 68022207-68022207 on Build GRCh38
rs754371841 Chr.16: 68022124-68022124 on Build GRCh38
rs757167665 Chr.16: 68022197-68022197 on Build GRCh38
rs1013985028 Chr.16: 68022117-68022117 on Build GRCh38
rs777986879 Chr.16: 68022185-68022185 on Build GRCh38
rs908517081 Chr.16: 68022214-68022214 on Build GRCh38
rs149411398 Chr.16: 68022084-68022084 on Build GRCh38
rs765568465 Chr.16: 68022143-68022143 on Build GRCh38
rs374594945 Chr.16: 68022230-68022230 on Build GRCh38
rs751802444 Chr.16: 68022165-68022165 on Build GRCh38
rs200156577 Chr.16: 68022112-68022112 on Build GRCh38
rs766801673 Chr.16: 68022116-68022116 on Build GRCh38
rs780154620 Chr.16: 68022065-68022065 on Build GRCh38
rs780103133 Chr.16: 68022196-68022196 on Build GRCh38
rs934219910 Chr.16: 68022195-68022195 on Build GRCh38
rs200912601 Chr.16: 68022190-68022190 on Build GRCh38
rs752931661 Chr.16: 68022147-68022147 on Build GRCh38
rs578218144 Chr.16: 68022218-68022218 on Build GRCh38
rs537364740 Chr.16: 68022224-68022224 on Build GRCh38
rs771584754 Chr.16: 68022100-68022100 on Build GRCh38
rs772852484 Chr.16: 68022232-68022232 on Build GRCh38
rs778618223 Chr.16: 68022222-68022222 on Build GRCh38
rs746480043 Chr.16: 68022233-68022233 on Build GRCh38
rs970190461 Chr.16: 68022174-68022174 on Build GRCh38
rs1023126661 Chr.16: 68022122-68022122 on Build GRCh38
rs760187645 Chr.16: 68022113-68022113 on Build GRCh38
rs570050909 Chr.16: 68022067-68022067 on Build GRCh38
rs759893892 Chr.16: 68022137-68022137 on Build GRCh38
rs147482514 Chr.16: 68022093-68022093 on Build GRCh38

View Full Product Details