Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52687411-52687658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNL3
Entrez Gene ID
26354
Gene Name
G protein nucleolar 3
Gene Aliases
C77032, E2IG3, NNP47, NS
UniGene
Hs.313544
Gene Chromosome Location
Chr.3: 52685920-52694497 on Build GRCh38
Transcripts
NM_014366.4 NM_206825.1 NM_206826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3596205 c.287C>T p.P96L Chr.3: 52687578-52687578 on Build GRCh38
6564224 c.234G>A p.Q78Q Chr.3: 52687525-52687525 on Build GRCh38
1047113 c.297G>A p.K99K Chr.3: 52687588-52687588 on Build GRCh38
323504 c.241G>C p.D81H Chr.3: 52687532-52687532 on Build GRCh38
1617881 c.268A>G p.R90G Chr.3: 52687559-52687559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767328590 Chr.3: 52687610-52687610 on Build GRCh38
rs754087813 Chr.3: 52687624-52687624 on Build GRCh38
rs781357099 Chr.3: 52687480-52687480 on Build GRCh38
rs779205225 Chr.3: 52687504-52687504 on Build GRCh38
rs750451380 Chr.3: 52687488-52687488 on Build GRCh38
rs369989113 Chr.3: 52687470-52687470 on Build GRCh38
rs757103419 Chr.3: 52687476-52687476 on Build GRCh38
rs750381380 Chr.3: 52687558-52687560 on Build GRCh38
rs756138949 Chr.3: 52687616-52687616 on Build GRCh38
rs199988647 Chr.3: 52687591-52687591 on Build GRCh38
rs34935779 Chr.3: 52687620-52687620 on Build GRCh38
rs761862028 Chr.3: 52687606-52687606 on Build GRCh38
rs756402309 Chr.3: 52687493-52687493 on Build GRCh38
rs758964197 Chr.3: 52687549-52687549 on Build GRCh38
rs765027651 Chr.3: 52687549-52687553 on Build GRCh38
rs138566929 Chr.3: 52687529-52687529 on Build GRCh38
rs776242992 Chr.3: 52687543-52687543 on Build GRCh38
rs973287570 Chr.3: 52687511-52687511 on Build GRCh38
rs142105228 Chr.3: 52687497-52687497 on Build GRCh38
rs775295433 Chr.3: 52687566-52687566 on Build GRCh38
rs762629052 Chr.3: 52687567-52687567 on Build GRCh38
rs780037109 Chr.3: 52687622-52687622 on Build GRCh38
rs200942271 Chr.3: 52687586-52687586 on Build GRCh38
rs764013060 Chr.3: 52687582-52687582 on Build GRCh38
rs762694522 Chr.3: 52687477-52687477 on Build GRCh38
rs1014579701 Chr.3: 52687466-52687466 on Build GRCh38
rs750586902 Chr.3: 52687613-52687613 on Build GRCh38
rs769009954 Chr.3: 52687503-52687503 on Build GRCh38
rs749450489 Chr.3: 52687500-52687500 on Build GRCh38
rs201521320 Chr.3: 52687540-52687540 on Build GRCh38
rs758090017 Chr.3: 52687630-52687631 on Build GRCh38
rs769627492 Chr.3: 52687551-52687551 on Build GRCh38
rs758255101 Chr.3: 52687451-52687451 on Build GRCh38
rs763924869 Chr.3: 52687459-52687459 on Build GRCh38
rs373178553 Chr.3: 52687452-52687452 on Build GRCh38
rs375780931 Chr.3: 52687490-52687490 on Build GRCh38
rs772434267 Chr.3: 52687533-52687533 on Build GRCh38

View Full Product Details