Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1853002-1853275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEIOB
Entrez Gene ID
254528
Gene Name
meiosis specific with OB domains
Gene Aliases
C16orf73, gs129
UniGene
Hs.729604
Gene Chromosome Location
Chr.16: 1833983-1872178 on Build GRCh38
Transcripts
NM_001163560.2 NM_152764.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6256241 c.670C>A p.P224T Chr.16: 1853231-1853231 on Build GRCh38
5054692 c.699T>C p.D233D Chr.16: 1853118-1853118 on Build GRCh38
7376911 c.28A>G p.I10V Chr.16: 1853057-1853057 on Build GRCh38
4994675 c.680C>A p.T227K Chr.16: 1853221-1853221 on Build GRCh38
5946661 c.717C>A p.D239E Chr.16: 1853100-1853100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771778959 Chr.16: 1853204-1853204 on Build GRCh38
rs779895795 Chr.16: 1853141-1853141 on Build GRCh38
rs1026337175 Chr.16: 1853120-1853120 on Build GRCh38
rs775787759 Chr.16: 1853100-1853100 on Build GRCh38
rs770295523 Chr.16: 1853093-1853093 on Build GRCh38
rs760422538 Chr.16: 1853030-1853031 on Build GRCh38
rs750376392 Chr.16: 1853135-1853135 on Build GRCh38
rs768617760 Chr.16: 1853097-1853098 on Build GRCh38
rs763333784 Chr.16: 1853117-1853117 on Build GRCh38
rs938577067 Chr.16: 1853183-1853183 on Build GRCh38
rs747656917 Chr.16: 1853205-1853205 on Build GRCh38
rs1057094122 Chr.16: 1853238-1853238 on Build GRCh38
rs778543387 Chr.16: 1853078-1853078 on Build GRCh38
rs777310422 Chr.16: 1853091-1853091 on Build GRCh38
rs768050594 Chr.16: 1853069-1853069 on Build GRCh38
rs761621564 Chr.16: 1853122-1853122 on Build GRCh38
rs1009403775 Chr.16: 1853123-1853123 on Build GRCh38
rs377077737 Chr.16: 1853055-1853055 on Build GRCh38
rs750255985 Chr.16: 1853057-1853057 on Build GRCh38
rs764238235 Chr.16: 1853047-1853047 on Build GRCh38
rs751523485 Chr.16: 1853190-1853191 on Build GRCh38
rs764062771 Chr.16: 1853118-1853118 on Build GRCh38
rs748947281 Chr.16: 1853067-1853067 on Build GRCh38
rs1045077718 Chr.16: 1853086-1853086 on Build GRCh38
rs372255449 Chr.16: 1853228-1853228 on Build GRCh38
rs370304721 Chr.16: 1853109-1853109 on Build GRCh38
rs987045080 Chr.16: 1853108-1853108 on Build GRCh38
rs766439374 Chr.16: 1853144-1853144 on Build GRCh38
rs879105371 Chr.16: 1853113-1853113 on Build GRCh38
rs755928794 Chr.16: 1853060-1853060 on Build GRCh38
rs201399419 Chr.16: 1853121-1853121 on Build GRCh38
rs767446432 Chr.16: 1853124-1853124 on Build GRCh38
rs759822119 Chr.16: 1853200-1853201 on Build GRCh38
rs372121011 Chr.16: 1853048-1853048 on Build GRCh38
rs201436120 Chr.16: 1853153-1853153 on Build GRCh38
rs372655502 Chr.16: 1853092-1853092 on Build GRCh38
rs76370825 Chr.16: 1853137-1853137 on Build GRCh38
rs778256765 Chr.16: 1853192-1853192 on Build GRCh38
rs373682180 Chr.16: 1853063-1853063 on Build GRCh38
rs747451643 Chr.16: 1853084-1853084 on Build GRCh38
rs557605700 Chr.16: 1853177-1853177 on Build GRCh38
rs975264511 Chr.16: 1853208-1853208 on Build GRCh38
rs776525949 Chr.16: 1853140-1853141 on Build GRCh38
rs763075708 Chr.16: 1853143-1853144 on Build GRCh38
rs541264555 Chr.16: 1853227-1853227 on Build GRCh38
rs202153809 Chr.16: 1853049-1853049 on Build GRCh38
rs568666095 Chr.16: 1853144-1853144 on Build GRCh38

View Full Product Details