Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24139179-24139446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC9
Entrez Gene ID
51016
Gene Name
ER membrane protein complex subunit 9
Gene Aliases
C14orf122, CGI-112, FAM158A
UniGene
Hs.271614
Gene Chromosome Location
Chr.14: 24138963-24142446 on Build GRCh38
Transcripts
NM_016049.3 XM_005267721.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4493422 c.414C>T p.L138L Chr.14: 24139386-24139386 on Build GRCh38
4826611 c.454G>A p.D152N Chr.14: 24139183-24139183 on Build GRCh38
4493288 c.412C>T p.L138F Chr.14: 24139388-24139388 on Build GRCh38
6653643 c.394G>A p.V132I Chr.14: 24139406-24139406 on Build GRCh38
7352607 c.376C>T p.R126C Chr.14: 24139424-24139424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556335154 Chr.14: 24139298-24139298 on Build GRCh38
rs143770220 Chr.14: 24139424-24139424 on Build GRCh38
rs374570620 Chr.14: 24139239-24139239 on Build GRCh38
rs771693003 Chr.14: 24139364-24139364 on Build GRCh38
rs950192200 Chr.14: 24139307-24139307 on Build GRCh38
rs781374678 Chr.14: 24139340-24139340 on Build GRCh38
rs374244627 Chr.14: 24139204-24139204 on Build GRCh38
rs574914404 Chr.14: 24139348-24139348 on Build GRCh38
rs1045126183 Chr.14: 24139312-24139312 on Build GRCh38
rs745827393 Chr.14: 24139350-24139350 on Build GRCh38
rs985204245 Chr.14: 24139203-24139203 on Build GRCh38
rs370804813 Chr.14: 24139237-24139237 on Build GRCh38
rs766614035 Chr.14: 24139403-24139403 on Build GRCh38
rs200649921 Chr.14: 24139215-24139215 on Build GRCh38
rs373329454 Chr.14: 24139414-24139414 on Build GRCh38
rs763552530 Chr.14: 24139228-24139228 on Build GRCh38
rs1009478129 Chr.14: 24139383-24139383 on Build GRCh38
rs765706144 Chr.14: 24139238-24139238 on Build GRCh38
rs780820560 Chr.14: 24139388-24139388 on Build GRCh38
rs778512392 Chr.14: 24139245-24139246 on Build GRCh38
rs753366011 Chr.14: 24139236-24139236 on Build GRCh38
rs149399035 Chr.14: 24139413-24139413 on Build GRCh38
rs755218563 Chr.14: 24139407-24139407 on Build GRCh38
rs576358950 Chr.14: 24139303-24139303 on Build GRCh38
rs377411623 Chr.14: 24139266-24139266 on Build GRCh38
rs760269356 Chr.14: 24139369-24139369 on Build GRCh38
rs751996416 Chr.14: 24139406-24139406 on Build GRCh38
rs775384061 Chr.14: 24139366-24139366 on Build GRCh38
rs201407803 Chr.14: 24139384-24139384 on Build GRCh38
rs774212286 Chr.14: 24139349-24139349 on Build GRCh38
rs887885961 Chr.14: 24139314-24139314 on Build GRCh38
rs201100583 Chr.14: 24139426-24139426 on Build GRCh38
rs758949552 Chr.14: 24139400-24139400 on Build GRCh38
rs763787218 Chr.14: 24139377-24139377 on Build GRCh38
rs748277594 Chr.14: 24139347-24139347 on Build GRCh38
rs896445167 Chr.14: 24139283-24139283 on Build GRCh38
rs776091070 Chr.14: 24139380-24139380 on Build GRCh38
rs184976129 Chr.14: 24139310-24139310 on Build GRCh38
rs373811642 Chr.14: 24139385-24139385 on Build GRCh38
rs200141151 Chr.14: 24139331-24139331 on Build GRCh38
rs772065541 Chr.14: 24139427-24139427 on Build GRCh38
rs371231907 Chr.14: 24139343-24139343 on Build GRCh38
rs923852680 Chr.14: 24139256-24139256 on Build GRCh38
rs756205254 Chr.14: 24139423-24139423 on Build GRCh38
rs755284078 Chr.14: 24139339-24139339 on Build GRCh38
rs554985462 Chr.14: 24139311-24139311 on Build GRCh38
rs747134681 Chr.14: 24139318-24139318 on Build GRCh38
rs775553452 Chr.14: 24139261-24139261 on Build GRCh38
rs1037033189 Chr.14: 24139279-24139279 on Build GRCh38

View Full Product Details