Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160293059-160293320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPA
Entrez Gene ID
1314
Gene Name
coatomer protein complex subunit alpha
Gene Aliases
AILJK, HEP-COP
UniGene
Hs.162121
Gene Chromosome Location
Chr.1: 160288587-160343564 on Build GRCh38
Transcripts
NM_001098398.1 NM_004371.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6344225 c.2825C>A p.A942E Chr.1: 160293191-160293191 on Build GRCh38
1295334 c.2850+1G>A p.? Chr.1: 160293165-160293165 on Build GRCh38
6344226 c.2798C>A p.A933E Chr.1: 160293191-160293191 on Build GRCh38
3476820 c.2823G>A p.R941R Chr.1: 160293166-160293166 on Build GRCh38
5542781 c.2804C>T p.S935F Chr.1: 160293185-160293185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753139776 Chr.1: 160293254-160293254 on Build GRCh38
rs935647535 Chr.1: 160293088-160293088 on Build GRCh38
rs754907028 Chr.1: 160293254-160293254 on Build GRCh38
rs760354732 Chr.1: 160293181-160293181 on Build GRCh38
rs370138103 Chr.1: 160293116-160293116 on Build GRCh38
rs765949860 Chr.1: 160293253-160293254 on Build GRCh38
rs756686942 Chr.1: 160293157-160293157 on Build GRCh38
rs879146777 Chr.1: 160293103-160293103 on Build GRCh38
rs749893916 Chr.1: 160293205-160293205 on Build GRCh38
rs750016153 Chr.1: 160293123-160293123 on Build GRCh38
rs367778679 Chr.1: 160293256-160293256 on Build GRCh38
rs781665205 Chr.1: 160293138-160293138 on Build GRCh38
rs767550206 Chr.1: 160293258-160293261 on Build GRCh38
rs751550523 Chr.1: 160293254-160293255 on Build GRCh38
rs776499023 Chr.1: 160293196-160293196 on Build GRCh38
rs969842258 Chr.1: 160293277-160293277 on Build GRCh38
rs764048827 Chr.1: 160293182-160293182 on Build GRCh38
rs765920287 Chr.1: 160293227-160293227 on Build GRCh38
rs200318072 Chr.1: 160293168-160293168 on Build GRCh38
rs552778606 Chr.1: 160293180-160293180 on Build GRCh38
rs748443010 Chr.1: 160293151-160293151 on Build GRCh38
rs764656530 Chr.1: 160293200-160293200 on Build GRCh38
rs150910743 Chr.1: 160293177-160293177 on Build GRCh38
rs778388338 Chr.1: 160293163-160293163 on Build GRCh38
rs12090071 Chr.1: 160293117-160293117 on Build GRCh38
rs780735416 Chr.1: 160293259-160293259 on Build GRCh38
rs367961082 Chr.1: 160293125-160293125 on Build GRCh38
rs761577917 Chr.1: 160293214-160293214 on Build GRCh38
rs368690601 Chr.1: 160293167-160293167 on Build GRCh38
rs778439520 Chr.1: 160293258-160293258 on Build GRCh38
rs372660936 Chr.1: 160293198-160293198 on Build GRCh38
rs749747568 Chr.1: 160293273-160293273 on Build GRCh38
rs560610472 Chr.1: 160293119-160293119 on Build GRCh38
rs1023754318 Chr.1: 160293261-160293261 on Build GRCh38
rs530092667 Chr.1: 160293111-160293111 on Build GRCh38

View Full Product Details