Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 169729553-169729810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPF1
Entrez Gene ID
54969
Gene Name
histone PARylation factor 1
Gene Aliases
C4orf27
UniGene
Hs.406756
Gene Chromosome Location
Chr.4: 169729468-169757954 on Build GRCh38
Transcripts
NM_017867.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1195898 c.1034C>T p.A345V Chr.4: 169729585-169729585 on Build GRCh38
1053196 c.929G>A p.G310D Chr.4: 169729690-169729690 on Build GRCh38
6327098 c.1012C>A p.Q338K Chr.4: 169729607-169729607 on Build GRCh38
6469407 c.1007G>A p.R336K Chr.4: 169729612-169729612 on Build GRCh38
3602023 c.951T>C p.Y317Y Chr.4: 169729668-169729668 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773864808 Chr.4: 169729697-169729697 on Build GRCh38
rs769414896 Chr.4: 169729680-169729681 on Build GRCh38
rs201314349 Chr.4: 169729609-169729609 on Build GRCh38
rs201211357 Chr.4: 169729580-169729580 on Build GRCh38
rs748544316 Chr.4: 169729672-169729672 on Build GRCh38
rs772197966 Chr.4: 169729636-169729636 on Build GRCh38
rs776407985 Chr.4: 169729666-169729666 on Build GRCh38
rs776936278 Chr.4: 169729635-169729635 on Build GRCh38
rs768640113 Chr.4: 169729652-169729653 on Build GRCh38
rs762671151 Chr.4: 169729713-169729718 on Build GRCh38
rs375900595 Chr.4: 169729702-169729702 on Build GRCh38
rs144533060 Chr.4: 169729616-169729616 on Build GRCh38
rs767103397 Chr.4: 169729730-169729730 on Build GRCh38
rs1011849896 Chr.4: 169729604-169729604 on Build GRCh38
rs768882243 Chr.4: 169729647-169729647 on Build GRCh38
rs752488911 Chr.4: 169729745-169729745 on Build GRCh38
rs1047716 Chr.4: 169729608-169729608 on Build GRCh38
rs557638387 Chr.4: 169729685-169729685 on Build GRCh38
rs146654985 Chr.4: 169729619-169729619 on Build GRCh38
rs956574202 Chr.4: 169729643-169729646 on Build GRCh38
rs759809862 Chr.4: 169729671-169729671 on Build GRCh38
rs368686466 Chr.4: 169729617-169729617 on Build GRCh38
rs975400402 Chr.4: 169729764-169729764 on Build GRCh38
rs912504388 Chr.4: 169729704-169729704 on Build GRCh38
rs765985756 Chr.4: 169729696-169729696 on Build GRCh38
rs1031214140 Chr.4: 169729628-169729628 on Build GRCh38
rs1047148101 Chr.4: 169729772-169729772 on Build GRCh38
rs1047726 Chr.4: 169729589-169729589 on Build GRCh38
rs772811309 Chr.4: 169729710-169729711 on Build GRCh38
rs762385457 Chr.4: 169729692-169729692 on Build GRCh38
rs773726375 Chr.4: 169729691-169729691 on Build GRCh38
rs370241692 Chr.4: 169729767-169729767 on Build GRCh38
rs780133428 Chr.4: 169729643-169729643 on Build GRCh38
rs747312133 Chr.4: 169729644-169729644 on Build GRCh38
rs762210746 Chr.4: 169729611-169729612 on Build GRCh38
rs374474538 Chr.4: 169729605-169729605 on Build GRCh38
rs754728516 Chr.4: 169729611-169729611 on Build GRCh38
rs747978215 Chr.4: 169729681-169729681 on Build GRCh38
rs1047706 Chr.4: 169729626-169729626 on Build GRCh38
rs1047719 Chr.4: 169729607-169729607 on Build GRCh38

View Full Product Details