Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26184699-26184928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM2
Entrez Gene ID
150946
Gene Name
GRB2 associated regulator of MAPK1 subtype 2
Gene Aliases
FAM59B, GAREML
UniGene
Hs.187912
Gene Chromosome Location
Chr.2: 26173091-26202405 on Build GRCh38
Transcripts
NM_001168241.1 NM_001191033.1 XM_006711951.3 XM_011532564.2 XM_011532565.2 XM_011532566.2 XM_011532567.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6031937 c.876T>C p.V292V Chr.2: 26184724-26184724 on Build GRCh38
6509166 c.972C>A p.F324L Chr.2: 26184820-26184820 on Build GRCh38
4952427 c.989T>C p.L330P Chr.2: 26184837-26184837 on Build GRCh38
5569475 c.893C>T p.T298M Chr.2: 26184741-26184741 on Build GRCh38
6841246 c.1006C>T p.R336C Chr.2: 26184854-26184854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917575464 Chr.2: 26184853-26184853 on Build GRCh38
rs868834620 Chr.2: 26184856-26184856 on Build GRCh38
rs761750999 Chr.2: 26184908-26184908 on Build GRCh38
rs771439596 Chr.2: 26184731-26184731 on Build GRCh38
rs746879943 Chr.2: 26184784-26184784 on Build GRCh38
rs530714572 Chr.2: 26184757-26184757 on Build GRCh38
rs945026258 Chr.2: 26184866-26184866 on Build GRCh38
rs1033962931 Chr.2: 26184779-26184779 on Build GRCh38
rs978278530 Chr.2: 26184892-26184892 on Build GRCh38
rs867251395 Chr.2: 26184882-26184882 on Build GRCh38
rs775083409 Chr.2: 26184777-26184777 on Build GRCh38
rs768784560 Chr.2: 26184805-26184805 on Build GRCh38
rs925053279 Chr.2: 26184895-26184895 on Build GRCh38
rs776557870 Chr.2: 26184820-26184820 on Build GRCh38
rs751025777 Chr.2: 26184858-26184858 on Build GRCh38
rs868387397 Chr.2: 26184880-26184880 on Build GRCh38

View Full Product Details