Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36469033-36469306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF3R
Entrez Gene ID
1441
Gene Name
colony stimulating factor 3 receptor
Gene Aliases
CD114, GCSFR, SCN7
UniGene
Hs.524517
Gene Chromosome Location
Chr.1: 36466043-36483314 on Build GRCh38
Transcripts
NM_000760.3 NM_156039.3 NM_172313.2 XM_005270493.1 XM_011540748.2 XM_011540749.1 XM_011540750.1 XM_017000370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6971444 c.1540C>T p.P514S Chr.1: 36469192-36469192 on Build GRCh38
908919 c.1542C>T p.P514P Chr.1: 36469190-36469190 on Build GRCh38
1748389 c.1480A>G p.I494V Chr.1: 36469252-36469252 on Build GRCh38
6952599 c.1501G>A p.E501K Chr.1: 36469231-36469231 on Build GRCh38
7136756 c.1552G>A p.V518I Chr.1: 36469180-36469180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906675527 Chr.1: 36469105-36469105 on Build GRCh38
rs566320564 Chr.1: 36469065-36469065 on Build GRCh38
rs922696012 Chr.1: 36469141-36469141 on Build GRCh38
rs149081693 Chr.1: 36469157-36469157 on Build GRCh38
rs776028338 Chr.1: 36469229-36469229 on Build GRCh38
rs772856183 Chr.1: 36469283-36469283 on Build GRCh38
rs777383194 Chr.1: 36469202-36469202 on Build GRCh38
rs764807760 Chr.1: 36469274-36469274 on Build GRCh38
rs772763576 Chr.1: 36469109-36469109 on Build GRCh38
rs747496320 Chr.1: 36469247-36469247 on Build GRCh38
rs142503546 Chr.1: 36469223-36469223 on Build GRCh38
rs761491589 Chr.1: 36469136-36469136 on Build GRCh38
rs572901231 Chr.1: 36469182-36469182 on Build GRCh38
rs377381632 Chr.1: 36469189-36469189 on Build GRCh38
rs929797975 Chr.1: 36469146-36469146 on Build GRCh38
rs34574943 Chr.1: 36469214-36469214 on Build GRCh38
rs1045435920 Chr.1: 36469253-36469253 on Build GRCh38
rs373426756 Chr.1: 36469121-36469121 on Build GRCh38
rs757856662 Chr.1: 36469144-36469144 on Build GRCh38
rs760117125 Chr.1: 36469106-36469106 on Build GRCh38
rs1018279235 Chr.1: 36469070-36469070 on Build GRCh38
rs551488904 Chr.1: 36469060-36469060 on Build GRCh38
rs1046580783 Chr.1: 36469072-36469072 on Build GRCh38
rs964068546 Chr.1: 36469078-36469078 on Build GRCh38
rs764719296 Chr.1: 36469139-36469139 on Build GRCh38
rs769045336 Chr.1: 36469248-36469248 on Build GRCh38
rs779124102 Chr.1: 36469217-36469217 on Build GRCh38
rs761438236 Chr.1: 36469270-36469270 on Build GRCh38
rs757913112 Chr.1: 36469162-36469162 on Build GRCh38
rs1031224658 Chr.1: 36469155-36469155 on Build GRCh38
rs3917991 Chr.1: 36469204-36469204 on Build GRCh38
rs2275472 Chr.1: 36469066-36469066 on Build GRCh38
rs548853956 Chr.1: 36469086-36469086 on Build GRCh38
rs775879363 Chr.1: 36469113-36469113 on Build GRCh38
rs776300248 Chr.1: 36469128-36469128 on Build GRCh38
rs766272961 Chr.1: 36469175-36469175 on Build GRCh38
rs145256322 Chr.1: 36469222-36469222 on Build GRCh38
rs761038239 Chr.1: 36469118-36469118 on Build GRCh38
rs375019697 Chr.1: 36469267-36469267 on Build GRCh38

View Full Product Details