Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7015697-7015954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf49
Entrez Gene ID
124944
Gene Name
chromosome 17 open reading frame 49
Gene Aliases
BAP18, HEPIS
UniGene
Hs.511801
Gene Chromosome Location
Chr.17: 7014737-7017524 on Build GRCh38
Transcripts
NM_001142798.2 NM_001142799.2 NM_174893.3 NR_038310.1 NR_037717.1

Gene Symbol
MIR497HG
Entrez Gene ID
100506755
Gene Name
mir-497-195 cluster host gene
Gene Aliases
MIR195HG
UniGene
Hs.710442
Gene Chromosome Location
Chr.17: 7015818-7019654 on Build GRCh38
Transcripts
NM_001142798.2 NM_001142799.2 NM_174893.3 NR_038310.1 NR_037717.1

Gene Symbol
RNASEK-C17orf49
Entrez Gene ID
100529209
Gene Name
RNASEK-C17orf49 readthrough
Gene Aliases
-
UniGene
Hs.511801
Gene Chromosome Location
Chr.17: 7012417-7017524 on Build GRCh38
Transcripts
NM_001142798.2 NM_001142799.2 NM_174893.3 NR_038310.1 NR_037717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4914091 c.218T>C p.V73A Chr.17: 7015875-7015875 on Build GRCh38
1750273 c.166C>T p.R56* Chr.17: 7015823-7015823 on Build GRCh38
4667051 c.178G>A p.D60N Chr.17: 7015835-7015835 on Build GRCh38
5438931 c.129G>A p.T43T Chr.17: 7015786-7015786 on Build GRCh38
349424 c.150G>A p.L50L Chr.17: 7015807-7015807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571259364 Chr.17: 7015807-7015807 on Build GRCh38
rs757619671 Chr.17: 7015741-7015741 on Build GRCh38
rs760188230 Chr.17: 7015735-7015735 on Build GRCh38
rs1048664169 Chr.17: 7015792-7015792 on Build GRCh38
rs778777537 Chr.17: 7015901-7015901 on Build GRCh38
rs765745746 Chr.17: 7015843-7015843 on Build GRCh38
rs1017472351 Chr.17: 7015751-7015752 on Build GRCh38
rs771739327 Chr.17: 7015800-7015800 on Build GRCh38
rs747090465 Chr.17: 7015763-7015763 on Build GRCh38
rs768089481 Chr.17: 7015926-7015926 on Build GRCh38
rs762350012 Chr.17: 7015842-7015842 on Build GRCh38
rs370840986 Chr.17: 7015788-7015788 on Build GRCh38
rs750849120 Chr.17: 7015852-7015852 on Build GRCh38
rs975313736 Chr.17: 7015777-7015777 on Build GRCh38
rs754141726 Chr.17: 7015729-7015730 on Build GRCh38
rs146139652 Chr.17: 7015858-7015858 on Build GRCh38
rs777632012 Chr.17: 7015771-7015771 on Build GRCh38
rs374433605 Chr.17: 7015754-7015754 on Build GRCh38
rs374069962 Chr.17: 7015756-7015756 on Build GRCh38
rs377766885 Chr.17: 7015761-7015761 on Build GRCh38
rs764510575 Chr.17: 7015736-7015736 on Build GRCh38
rs757445772 Chr.17: 7015730-7015752 on Build GRCh38
rs775053991 Chr.17: 7015805-7015805 on Build GRCh38
rs771794341 Chr.17: 7015912-7015912 on Build GRCh38
rs778731478 Chr.17: 7015730-7015730 on Build GRCh38
rs749236044 Chr.17: 7015885-7015885 on Build GRCh38
rs781303846 Chr.17: 7015869-7015869 on Build GRCh38
rs780438514 Chr.17: 7015860-7015860 on Build GRCh38
rs754311157 Chr.17: 7015737-7015737 on Build GRCh38
rs770674909 Chr.17: 7015786-7015786 on Build GRCh38
rs767067258 Chr.17: 7015727-7015727 on Build GRCh38
rs779331061 Chr.17: 7015742-7015742 on Build GRCh38
rs896184459 Chr.17: 7015729-7015729 on Build GRCh38
rs745660441 Chr.17: 7015757-7015758 on Build GRCh38
rs14309 Chr.17: 7015774-7015774 on Build GRCh38
rs1001987045 Chr.17: 7015720-7015720 on Build GRCh38
rs754368807 Chr.17: 7015833-7015833 on Build GRCh38
rs758671534 Chr.17: 7015750-7015750 on Build GRCh38
rs73237593 Chr.17: 7015733-7015733 on Build GRCh38
rs772073920 Chr.17: 7015884-7015904 on Build GRCh38
rs774114363 Chr.17: 7015787-7015787 on Build GRCh38
rs746667599 Chr.17: 7015916-7015916 on Build GRCh38
rs759161458 Chr.17: 7015723-7015723 on Build GRCh38
rs1530377 Chr.17: 7015913-7015913 on Build GRCh38
rs536953637 Chr.17: 7015863-7015863 on Build GRCh38
rs532408681 Chr.17: 7015850-7015850 on Build GRCh38
rs761245450 Chr.17: 7015815-7015815 on Build GRCh38
rs1044917 Chr.17: 7015896-7015896 on Build GRCh38
rs140134459 Chr.17: 7015868-7015868 on Build GRCh38
rs750714108 Chr.17: 7015747-7015747 on Build GRCh38
rs376403849 Chr.17: 7015904-7015904 on Build GRCh38
rs536473581 Chr.17: 7015931-7015931 on Build GRCh38
rs375220513 Chr.17: 7015827-7015827 on Build GRCh38
rs768724586 Chr.17: 7015765-7015765 on Build GRCh38

View Full Product Details