Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57621513-57621766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCOM1
Entrez Gene ID
145781
Gene Name
GRINL1A complex locus 1
Gene Aliases
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57717557 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Gene Symbol
MYZAP
Entrez Gene ID
100820829
Gene Name
myocardial zonula adherens protein
Gene Aliases
GCOM1, Gup, MYOZAP
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57685364 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018100.4 NM_152451.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5585874 c.370G>A p.E124K Chr.15: 57621659-57621659 on Build GRCh38
3502699 c.410C>T p.S137L Chr.15: 57621699-57621699 on Build GRCh38
3502697 c.410C>G p.S137* Chr.15: 57621699-57621699 on Build GRCh38
3502695 c.377G>A p.R126K Chr.15: 57621666-57621666 on Build GRCh38
1373743 c.361G>A p.D121N Chr.15: 57621650-57621650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021888770 Chr.15: 57621597-57621597 on Build GRCh38
rs551718913 Chr.15: 57621643-57621643 on Build GRCh38
rs753880816 Chr.15: 57621692-57621692 on Build GRCh38
rs758650024 Chr.15: 57621582-57621585 on Build GRCh38
rs759093310 Chr.15: 57621586-57621586 on Build GRCh38
rs368474764 Chr.15: 57621564-57621564 on Build GRCh38
rs760226520 Chr.15: 57621680-57621680 on Build GRCh38
rs551885647 Chr.15: 57621655-57621655 on Build GRCh38
rs199650252 Chr.15: 57621695-57621695 on Build GRCh38
rs925989191 Chr.15: 57621721-57621721 on Build GRCh38
rs749809839 Chr.15: 57621741-57621741 on Build GRCh38
rs748202049 Chr.15: 57621638-57621638 on Build GRCh38
rs770449271 Chr.15: 57621639-57621639 on Build GRCh38
rs774115646 Chr.15: 57621585-57621585 on Build GRCh38
rs755271606 Chr.15: 57621715-57621715 on Build GRCh38
rs750126404 Chr.15: 57621701-57621701 on Build GRCh38
rs773747314 Chr.15: 57621578-57621578 on Build GRCh38
rs369885749 Chr.15: 57621678-57621678 on Build GRCh38
rs747167606 Chr.15: 57621566-57621566 on Build GRCh38
rs367806616 Chr.15: 57621569-57621569 on Build GRCh38
rs866893705 Chr.15: 57621699-57621699 on Build GRCh38
rs752134567 Chr.15: 57621589-57621589 on Build GRCh38
rs998772664 Chr.15: 57621640-57621640 on Build GRCh38
rs777714126 Chr.15: 57621738-57621738 on Build GRCh38
rs778919534 Chr.15: 57621626-57621626 on Build GRCh38
rs200416831 Chr.15: 57621577-57621577 on Build GRCh38
rs771380707 Chr.15: 57621652-57621652 on Build GRCh38
rs747315874 Chr.15: 57621616-57621616 on Build GRCh38
rs375289402 Chr.15: 57621677-57621677 on Build GRCh38
rs57874684 Chr.15: 57621602-57621602 on Build GRCh38
rs763563004 Chr.15: 57621686-57621686 on Build GRCh38
rs752632343 Chr.15: 57621708-57621708 on Build GRCh38
rs757115112 Chr.15: 57621612-57621612 on Build GRCh38
rs768848968 Chr.15: 57621567-57621567 on Build GRCh38
rs773567348 Chr.15: 57621568-57621568 on Build GRCh38
rs375642380 Chr.15: 57621697-57621697 on Build GRCh38
rs778569189 Chr.15: 57621614-57621614 on Build GRCh38
rs766634581 Chr.15: 57621580-57621580 on Build GRCh38
rs956037395 Chr.15: 57621736-57621736 on Build GRCh38
rs372031185 Chr.15: 57621603-57621603 on Build GRCh38
rs756307516 Chr.15: 57621733-57621733 on Build GRCh38
rs755195995 Chr.15: 57621622-57621622 on Build GRCh38
rs752932553 Chr.15: 57621717-57621717 on Build GRCh38
rs375404503 Chr.15: 57621573-57621573 on Build GRCh38

View Full Product Details