Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117840268-117840515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD6
Entrez Gene ID
53826
Gene Name
FXYD domain containing ion transport regulator 6
Gene Aliases
-
UniGene
Hs.744850
Gene Chromosome Location
Chr.11: 117836976-117877486 on Build GRCh38
Transcripts
NM_001164831.2 NM_001164832.2 NM_001164836.2 NM_001164837.2 NM_022003.3 NM_001204268.1 NM_001243598.2

Gene Symbol
FXYD6-FXYD2
Entrez Gene ID
100533181
Gene Name
FXYD6-FXYD2 readthrough
Gene Aliases
-
UniGene
Hs.744850
Gene Chromosome Location
Chr.11: 117820075-117877031 on Build GRCh38
Transcripts
NM_001164831.2 NM_001164832.2 NM_001164836.2 NM_001164837.2 NM_022003.3 NM_001204268.1 NM_001243598.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3443844 c.235G>T p.V79L Chr.11: 117840343-117840343 on Build GRCh38
6067609 c.213C>A p.A71A Chr.11: 117840365-117840365 on Build GRCh38
6667998 c.252C>T p.T84T Chr.11: 117840326-117840326 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748096826 Chr.11: 117840405-117840405 on Build GRCh38
rs557049060 Chr.11: 117840418-117840445 on Build GRCh38
rs71482118 Chr.11: 117840377-117840377 on Build GRCh38
rs762999755 Chr.11: 117840295-117840295 on Build GRCh38
rs112003793 Chr.11: 117840370-117840370 on Build GRCh38
rs199942688 Chr.11: 117840308-117840308 on Build GRCh38
rs753233831 Chr.11: 117840309-117840309 on Build GRCh38
rs549725282 Chr.11: 117840461-117840461 on Build GRCh38
rs899158553 Chr.11: 117840425-117840425 on Build GRCh38
rs780911025 Chr.11: 117840386-117840386 on Build GRCh38
rs373937556 Chr.11: 117840321-117840321 on Build GRCh38
rs767958686 Chr.11: 117840429-117840429 on Build GRCh38
rs938835316 Chr.11: 117840319-117840319 on Build GRCh38
rs759910324 Chr.11: 117840307-117840307 on Build GRCh38
rs997167255 Chr.11: 117840494-117840494 on Build GRCh38
rs375394300 Chr.11: 117840325-117840326 on Build GRCh38
rs202139278 Chr.11: 117840325-117840325 on Build GRCh38
rs111727597 Chr.11: 117840431-117840431 on Build GRCh38
rs148930995 Chr.11: 117840474-117840474 on Build GRCh38
rs1008031393 Chr.11: 117840434-117840434 on Build GRCh38
rs775160010 Chr.11: 117840430-117840430 on Build GRCh38
rs757351154 Chr.11: 117840355-117840355 on Build GRCh38
rs773300077 Chr.11: 117840410-117840410 on Build GRCh38
rs753857831 Chr.11: 117840326-117840326 on Build GRCh38
rs768478976 Chr.11: 117840385-117840385 on Build GRCh38
rs141358460 Chr.11: 117840361-117840361 on Build GRCh38
rs916178909 Chr.11: 117840357-117840357 on Build GRCh38
rs769791951 Chr.11: 117840409-117840409 on Build GRCh38
rs1006026765 Chr.11: 117840413-117840413 on Build GRCh38

View Full Product Details