Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49665785-49666046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L12
Entrez Gene ID
83596
Gene Name
BCL2 like 12
Gene Aliases
-
UniGene
Hs.289052
Gene Chromosome Location
Chr.19: 49665072-49673916 on Build GRCh38
Transcripts
NM_001040668.1 NM_001282516.1 NM_001282517.1 NM_001282519.1 NM_001282520.1 NM_001282521.1 NM_138639.1 NR_104200.1 NR_104201.1 NR_104202.1 NR_104203.1 NR_104204.1 NR_104205.1 XM_017027345.1 XM_017027346.1 NM_001197122.1 NM_001197123.1 NM_001197124.1 NM_001197125.1 NM_001197126.1 NM_001197127.1 NM_001197128.1 NM_001571.5 NR_045568.1 XM_006723197.1 XM_006723198.1 XM_006723202.2 XM_017026766.1 XM_017026767.1

Gene Symbol
IRF3
Entrez Gene ID
3661
Gene Name
interferon regulatory factor 3
Gene Aliases
IIAE7
UniGene
Hs.289052
Gene Chromosome Location
Chr.19: 49659569-49665875 on Build GRCh38
Transcripts
NM_001040668.1 NM_001282516.1 NM_001282517.1 NM_001282519.1 NM_001282520.1 NM_001282521.1 NM_138639.1 NR_104200.1 NR_104201.1 NR_104202.1 NR_104203.1 NR_104204.1 NR_104205.1 XM_017027345.1 XM_017027346.1 NM_001197122.1 NM_001197123.1 NM_001197124.1 NM_001197125.1 NM_001197126.1 NM_001197127.1 NM_001197128.1 NM_001571.5 NR_045568.1 XM_006723197.1 XM_006723198.1 XM_006723202.2 XM_017026766.1 XM_017026767.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5546227 c.25C>T p.P9S Chr.19: 49665848-49665848 on Build GRCh38
5506538 c.116T>C p.L39P Chr.19: 49665939-49665939 on Build GRCh38
6610701 c.39delT p.L15fs*43 Chr.19: 49665862-49665862 on Build GRCh38
5542635 c.211G>A p.E71K Chr.19: 49666034-49666034 on Build GRCh38
3536896 c.19C>T p.L7L Chr.19: 49665842-49665842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777663072 Chr.19: 49665932-49665932 on Build GRCh38
rs370042754 Chr.19: 49665934-49665934 on Build GRCh38
rs142687869 Chr.19: 49665940-49665940 on Build GRCh38
rs922241817 Chr.19: 49666006-49666006 on Build GRCh38
rs753664304 Chr.19: 49665827-49665827 on Build GRCh38
rs761045724 Chr.19: 49666004-49666004 on Build GRCh38
rs775985642 Chr.19: 49665944-49665944 on Build GRCh38
rs755216778 Chr.19: 49665832-49665832 on Build GRCh38
rs746218557 Chr.19: 49665937-49665937 on Build GRCh38
rs778347169 Chr.19: 49665876-49665876 on Build GRCh38
rs564623657 Chr.19: 49665997-49665997 on Build GRCh38
rs776663973 Chr.19: 49665918-49665918 on Build GRCh38
rs902883580 Chr.19: 49665954-49665954 on Build GRCh38
rs141156787 Chr.19: 49665955-49665955 on Build GRCh38
rs148875442 Chr.19: 49665859-49665859 on Build GRCh38
rs771708612 Chr.19: 49665897-49665897 on Build GRCh38
rs761932915 Chr.19: 49665861-49665862 on Build GRCh38
rs150208335 Chr.19: 49665824-49665824 on Build GRCh38
rs778534728 Chr.19: 49665830-49665830 on Build GRCh38
rs760672189 Chr.19: 49665826-49665826 on Build GRCh38
rs771347725 Chr.19: 49665878-49665878 on Build GRCh38
rs199911047 Chr.19: 49665850-49665850 on Build GRCh38
rs761805340 Chr.19: 49666008-49666008 on Build GRCh38
rs768237927 Chr.19: 49665942-49665942 on Build GRCh38
rs766235924 Chr.19: 49665951-49665951 on Build GRCh38
rs138111341 Chr.19: 49665933-49665933 on Build GRCh38
rs779402883 Chr.19: 49665936-49665936 on Build GRCh38
rs757843109 Chr.19: 49665962-49665962 on Build GRCh38
rs762842795 Chr.19: 49665950-49665950 on Build GRCh38
rs145762717 Chr.19: 49665841-49665841 on Build GRCh38
rs748021413 Chr.19: 49665925-49665925 on Build GRCh38
rs748220773 Chr.19: 49665863-49665863 on Build GRCh38
rs373601090 Chr.19: 49666023-49666023 on Build GRCh38
rs1054305255 Chr.19: 49665961-49665961 on Build GRCh38
rs267605590 Chr.19: 49665831-49665831 on Build GRCh38
rs759274959 Chr.19: 49665879-49665879 on Build GRCh38
rs998085984 Chr.19: 49665845-49665845 on Build GRCh38
rs544779018 Chr.19: 49665992-49665992 on Build GRCh38
rs148185263 Chr.19: 49665949-49665949 on Build GRCh38
rs761874395 Chr.19: 49665945-49665945 on Build GRCh38
rs761609418 Chr.19: 49665919-49665919 on Build GRCh38
rs201895036 Chr.19: 49665970-49665970 on Build GRCh38
rs753065447 Chr.19: 49665915-49665915 on Build GRCh38
rs763532112 Chr.19: 49665911-49665911 on Build GRCh38
rs774082745 Chr.19: 49665818-49665818 on Build GRCh38
rs560898780 Chr.19: 49666018-49666018 on Build GRCh38
rs973664955 Chr.19: 49665900-49665900 on Build GRCh38
rs780440198 Chr.19: 49666027-49666027 on Build GRCh38
rs760195628 Chr.19: 49665910-49665910 on Build GRCh38
rs1029538760 Chr.19: 49665964-49665964 on Build GRCh38
rs909552758 Chr.19: 49665814-49665814 on Build GRCh38
rs779702428 Chr.19: 49665997-49665998 on Build GRCh38
rs376704095 Chr.19: 49665927-49665927 on Build GRCh38
rs763388210 Chr.19: 49665816-49665816 on Build GRCh38
rs144909468 Chr.19: 49665948-49665948 on Build GRCh38
rs776259017 Chr.19: 49665991-49665991 on Build GRCh38
rs750051880 Chr.19: 49665830-49665831 on Build GRCh38
rs749300966 Chr.19: 49665996-49665997 on Build GRCh38
rs200984195 Chr.19: 49665920-49665920 on Build GRCh38
rs772366601 Chr.19: 49665939-49665939 on Build GRCh38
rs145496469 Chr.19: 49665922-49665922 on Build GRCh38
rs759060438 Chr.19: 49665822-49665822 on Build GRCh38
rs1014988971 Chr.19: 49665990-49665990 on Build GRCh38
rs752989868 Chr.19: 49665953-49665953 on Build GRCh38
rs975100268 Chr.19: 49665996-49665996 on Build GRCh38
rs949868626 Chr.19: 49665952-49665952 on Build GRCh38
rs370560242 Chr.19: 49665994-49665994 on Build GRCh38
rs79741544 Chr.19: 49666007-49666007 on Build GRCh38
rs746717146 Chr.19: 49665980-49665980 on Build GRCh38
rs762932481 Chr.19: 49665995-49665995 on Build GRCh38
rs767861480 Chr.19: 49665923-49665923 on Build GRCh38
rs2060263 Chr.19: 49665963-49665963 on Build GRCh38
rs34958637 Chr.19: 49665944-49665945 on Build GRCh38
rs1009012618 Chr.19: 49665988-49665988 on Build GRCh38
rs750953700 Chr.19: 49666019-49666019 on Build GRCh38
rs78186439 Chr.19: 49665877-49665877 on Build GRCh38
rs752173647 Chr.19: 49665825-49665825 on Build GRCh38
rs267605591 Chr.19: 49665874-49665874 on Build GRCh38
rs775039915 Chr.19: 49665898-49665898 on Build GRCh38
rs774256421 Chr.19: 49666002-49666002 on Build GRCh38
rs756293294 Chr.19: 49665931-49665931 on Build GRCh38
rs780352220 Chr.19: 49665972-49665972 on Build GRCh38
rs567569942 Chr.19: 49665829-49665829 on Build GRCh38
rs768185508 Chr.19: 49665986-49665986 on Build GRCh38
rs771134637 Chr.19: 49665817-49665817 on Build GRCh38
rs772990009 Chr.19: 49665957-49665957 on Build GRCh38
rs150028589 Chr.19: 49665959-49665959 on Build GRCh38

View Full Product Details