Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 100129464-100129725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVL
Entrez Gene ID
51466
Gene Name
Enah/Vasp-like
Gene Aliases
RNB6
UniGene
Hs.125867
Gene Chromosome Location
Chr.14: 99971454-100144236 on Build GRCh38
Transcripts
NM_016337.2 XM_005267749.3 XM_011536828.2 XM_017021363.1 XM_017021364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4049054 c.726C>T p.D242D Chr.14: 100129571-100129571 on Build GRCh38
416290 c.776G>A p.R259Q Chr.14: 100129621-100129621 on Build GRCh38
4049055 c.738C>T p.G246G Chr.14: 100129583-100129583 on Build GRCh38
6489914 c.736G>A p.G246S Chr.14: 100129581-100129581 on Build GRCh38
32675 c.746C>T p.P249L Chr.14: 100129591-100129591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758161580 Chr.14: 100129707-100129707 on Build GRCh38
rs776343870 Chr.14: 100129645-100129645 on Build GRCh38
rs145112596 Chr.14: 100129571-100129571 on Build GRCh38
rs765988507 Chr.14: 100129640-100129640 on Build GRCh38
rs267604123 Chr.14: 100129622-100129622 on Build GRCh38
rs541896738 Chr.14: 100129542-100129542 on Build GRCh38
rs1036867242 Chr.14: 100129502-100129502 on Build GRCh38
rs144419602 Chr.14: 100129620-100129620 on Build GRCh38
rs748500160 Chr.14: 100129611-100129611 on Build GRCh38
rs758041364 Chr.14: 100129500-100129502 on Build GRCh38
rs566542060 Chr.14: 100129621-100129621 on Build GRCh38
rs774766360 Chr.14: 100129610-100129610 on Build GRCh38
rs200967615 Chr.14: 100129570-100129570 on Build GRCh38
rs925915240 Chr.14: 100129562-100129562 on Build GRCh38
rs764843531 Chr.14: 100129703-100129703 on Build GRCh38
rs200011657 Chr.14: 100129704-100129704 on Build GRCh38
rs368376778 Chr.14: 100129572-100129572 on Build GRCh38
rs777614006 Chr.14: 100129545-100129545 on Build GRCh38
rs553991967 Chr.14: 100129595-100129595 on Build GRCh38
rs751839633 Chr.14: 100129690-100129692 on Build GRCh38
rs919319412 Chr.14: 100129504-100129504 on Build GRCh38
rs74085121 Chr.14: 100129543-100129543 on Build GRCh38
rs990842872 Chr.14: 100129484-100129484 on Build GRCh38
rs889532442 Chr.14: 100129705-100129705 on Build GRCh38
rs758331510 Chr.14: 100129544-100129544 on Build GRCh38
rs374731942 Chr.14: 100129697-100129697 on Build GRCh38
rs766079655 Chr.14: 100129512-100129512 on Build GRCh38
rs754749586 Chr.14: 100129531-100129531 on Build GRCh38
rs768934680 Chr.14: 100129607-100129607 on Build GRCh38
rs140562245 Chr.14: 100129587-100129587 on Build GRCh38
rs779734142 Chr.14: 100129584-100129584 on Build GRCh38
rs753557503 Chr.14: 100129522-100129522 on Build GRCh38
rs112416267 Chr.14: 100129686-100129686 on Build GRCh38

View Full Product Details