Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176322156-176322427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIMC1
Entrez Gene ID
375484
Gene Name
SUMO interacting motifs containing 1
Gene Aliases
C5orf25, OOMA1, PLEIAD
UniGene
Hs.719847
Gene Chromosome Location
Chr.5: 176238359-176345991 on Build GRCh38
Transcripts
NM_001308195.1 NM_001308196.1 NM_001308200.1 NM_198567.5 NR_131772.1 XM_011534553.2 XM_011534554.2 XM_011534556.2 XM_017009454.1 XM_017009455.1 XM_017009456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
737657 c.1900C>A p.Q634K Chr.5: 176322340-176322340 on Build GRCh38
6326453 c.237C>T p.V79V Chr.5: 176322294-176322294 on Build GRCh38
1065988 c.1978A>G p.N660D Chr.5: 176322418-176322418 on Build GRCh38
5031404 c.1916C>T p.T639I Chr.5: 176322356-176322356 on Build GRCh38
6326452 c.1854C>T p.V618V Chr.5: 176322294-176322294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759347270 Chr.5: 176322275-176322275 on Build GRCh38
rs766231519 Chr.5: 176322212-176322212 on Build GRCh38
rs201575513 Chr.5: 176322345-176322345 on Build GRCh38
rs767509637 Chr.5: 176322280-176322280 on Build GRCh38
rs948394801 Chr.5: 176322371-176322371 on Build GRCh38
rs751083459 Chr.5: 176322265-176322265 on Build GRCh38
rs375596432 Chr.5: 176322301-176322301 on Build GRCh38
rs769023165 Chr.5: 176322331-176322331 on Build GRCh38
rs936844714 Chr.5: 176322285-176322285 on Build GRCh38
rs751364959 Chr.5: 176322298-176322298 on Build GRCh38
rs779960622 Chr.5: 176322328-176322328 on Build GRCh38
rs540631061 Chr.5: 176322229-176322229 on Build GRCh38
rs767057287 Chr.5: 176322369-176322369 on Build GRCh38
rs775138842 Chr.5: 176322384-176322391 on Build GRCh38
rs756044825 Chr.5: 176322293-176322293 on Build GRCh38
rs773577648 Chr.5: 176322352-176322352 on Build GRCh38
rs769576872 Chr.5: 176322257-176322258 on Build GRCh38
rs748869578 Chr.5: 176322391-176322391 on Build GRCh38
rs374074121 Chr.5: 176322281-176322281 on Build GRCh38
rs914340843 Chr.5: 176322339-176322339 on Build GRCh38
rs199680642 Chr.5: 176322253-176322253 on Build GRCh38
rs747134412 Chr.5: 176322329-176322329 on Build GRCh38
rs151327906 Chr.5: 176322294-176322294 on Build GRCh38
rs772914765 Chr.5: 176322242-176322242 on Build GRCh38
rs748507742 Chr.5: 176322342-176322342 on Build GRCh38
rs770644911 Chr.5: 176322401-176322401 on Build GRCh38
rs985885135 Chr.5: 176322185-176322185 on Build GRCh38
rs561718964 Chr.5: 176322335-176322335 on Build GRCh38
rs765746756 Chr.5: 176322264-176322264 on Build GRCh38
rs796958633 Chr.5: 176322212-176322213 on Build GRCh38
rs139666745 Chr.5: 176322366-176322366 on Build GRCh38
rs750618402 Chr.5: 176322296-176322296 on Build GRCh38
rs546994883 Chr.5: 176322233-176322236 on Build GRCh38
rs557974833 Chr.5: 176322235-176322235 on Build GRCh38

View Full Product Details