Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129613396-129613637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf50
Entrez Gene ID
375759
Gene Name
chromosome 9 open reading frame 50
Gene Aliases
-
UniGene
Hs.124223
Gene Chromosome Location
Chr.9: 129612225-129622275 on Build GRCh38
Transcripts
NM_199350.3 XM_011518655.1 XM_011518656.1 XM_011518657.2 XM_011518658.2 XM_011518659.1 XM_011518660.1 XM_011518661.2 XM_011518662.2 XM_011518663.1 XM_017014708.1 NM_001286796.1

Gene Symbol
NTMT1
Entrez Gene ID
28989
Gene Name
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Gene Aliases
AD-003, C9orf32, HOMT1A, METTL11A, NRMT, NTM1A
UniGene
Hs.744027
Gene Chromosome Location
Chr.9: 129608884-129636742 on Build GRCh38
Transcripts
NM_199350.3 XM_011518655.1 XM_011518656.1 XM_011518657.2 XM_011518658.2 XM_011518659.1 XM_011518660.1 XM_011518661.2 XM_011518662.2 XM_011518663.1 XM_017014708.1 NM_001286796.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1173452 c.964G>A p.E322K Chr.9: 129613514-129613514 on Build GRCh38
77402 c.914C>G p.A305G Chr.9: 129613564-129613564 on Build GRCh38
391978 c.910delG p.A304fs*60 Chr.9: 129613568-129613568 on Build GRCh38
3375121 c.1000G>A p.E334K Chr.9: 129613478-129613478 on Build GRCh38
4563536 c.979G>A p.A327T Chr.9: 129613499-129613499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746856775 Chr.9: 129613540-129613540 on Build GRCh38
rs968336638 Chr.9: 129613556-129613556 on Build GRCh38
rs370776156 Chr.9: 129613535-129613535 on Build GRCh38
rs752343580 Chr.9: 129613573-129613573 on Build GRCh38
rs757643011 Chr.9: 129613513-129613513 on Build GRCh38
rs749373438 Chr.9: 129613579-129613579 on Build GRCh38
rs757513239 Chr.9: 129613419-129613419 on Build GRCh38
rs149675106 Chr.9: 129613581-129613581 on Build GRCh38
rs138485391 Chr.9: 129613531-129613531 on Build GRCh38
rs373916566 Chr.9: 129613539-129613539 on Build GRCh38
rs779768131 Chr.9: 129613422-129613422 on Build GRCh38
rs759821940 Chr.9: 129613473-129613473 on Build GRCh38
rs745685264 Chr.9: 129613616-129613616 on Build GRCh38
rs762281716 Chr.9: 129613560-129613560 on Build GRCh38
rs150697927 Chr.9: 129613544-129613544 on Build GRCh38
rs781077632 Chr.9: 129613597-129613597 on Build GRCh38
rs770576697 Chr.9: 129613465-129613465 on Build GRCh38
rs763032856 Chr.9: 129613550-129613550 on Build GRCh38
rs139948871 Chr.9: 129613566-129613566 on Build GRCh38
rs756511872 Chr.9: 129613565-129613565 on Build GRCh38
rs751951762 Chr.9: 129613512-129613512 on Build GRCh38
rs752813624 Chr.9: 129613487-129613487 on Build GRCh38
rs374732466 Chr.9: 129613447-129613447 on Build GRCh38
rs143109321 Chr.9: 129613505-129613505 on Build GRCh38
rs764011501 Chr.9: 129613551-129613551 on Build GRCh38
rs750800254 Chr.9: 129613520-129613520 on Build GRCh38
rs751652325 Chr.9: 129613552-129613552 on Build GRCh38
rs36114608 Chr.9: 129613511-129613512 on Build GRCh38
rs41276772 Chr.9: 129613460-129613460 on Build GRCh38
rs547056212 Chr.9: 129613547-129613547 on Build GRCh38
rs781474219 Chr.9: 129613519-129613519 on Build GRCh38
rs750853346 Chr.9: 129613563-129613563 on Build GRCh38
rs370300951 Chr.9: 129613442-129613442 on Build GRCh38
rs376865697 Chr.9: 129613416-129613416 on Build GRCh38
rs762979793 Chr.9: 129613491-129613491 on Build GRCh38
rs147521610 Chr.9: 129613527-129613527 on Build GRCh38
rs1024080988 Chr.9: 129613567-129613567 on Build GRCh38
rs774059558 Chr.9: 129613455-129613455 on Build GRCh38
rs535635348 Chr.9: 129613586-129613586 on Build GRCh38
rs747635774 Chr.9: 129613456-129613456 on Build GRCh38
rs772180321 Chr.9: 129613459-129613459 on Build GRCh38
rs956519022 Chr.9: 129613582-129613582 on Build GRCh38
rs57250395 Chr.9: 129613417-129613417 on Build GRCh38
rs531243205 Chr.9: 129613468-129613468 on Build GRCh38
rs770823983 Chr.9: 129613589-129613589 on Build GRCh38
rs768415449 Chr.9: 129613443-129613443 on Build GRCh38
rs760155416 Chr.9: 129613576-129613576 on Build GRCh38
rs748602993 Chr.9: 129613526-129613526 on Build GRCh38
rs768041968 Chr.9: 129613562-129613562 on Build GRCh38
rs368683634 Chr.9: 129613608-129613608 on Build GRCh38
rs3213763 Chr.9: 129613543-129613543 on Build GRCh38
rs1039006654 Chr.9: 129613490-129613490 on Build GRCh38
rs146176082 Chr.9: 129613587-129613587 on Build GRCh38
rs566893379 Chr.9: 129613548-129613548 on Build GRCh38
rs368047969 Chr.9: 129613478-129613478 on Build GRCh38
rs956210101 Chr.9: 129613429-129613429 on Build GRCh38

View Full Product Details