Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34761803-34762050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJB4
Entrez Gene ID
127534
Gene Name
gap junction protein beta 4
Gene Aliases
CX30.3, EKV
UniGene
Hs.351203
Gene Chromosome Location
Chr.1: 34759741-34763724 on Build GRCh38
Transcripts
NM_153212.2 XM_011540679.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1342069 c.668G>A p.R223Q Chr.1: 34761922-34761922 on Build GRCh38
464475 c.678C>T p.C226C Chr.1: 34761932-34761932 on Build GRCh38
908575 c.746A>C p.N249T Chr.1: 34762000-34762000 on Build GRCh38
908574 c.724G>T p.G242* Chr.1: 34761978-34761978 on Build GRCh38
6126487 c.552G>T p.K184N Chr.1: 34761806-34761806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749875009 Chr.1: 34761923-34761923 on Build GRCh38
rs745548746 Chr.1: 34761887-34761887 on Build GRCh38
rs776398059 Chr.1: 34761901-34761901 on Build GRCh38
rs375489958 Chr.1: 34761928-34761928 on Build GRCh38
rs771162179 Chr.1: 34761906-34761906 on Build GRCh38
rs781088813 Chr.1: 34761941-34761941 on Build GRCh38
rs759845411 Chr.1: 34761908-34761908 on Build GRCh38
rs758125233 Chr.1: 34761927-34761927 on Build GRCh38
rs763878424 Chr.1: 34761838-34761838 on Build GRCh38
rs760944349 Chr.1: 34761913-34761913 on Build GRCh38
rs778397300 Chr.1: 34761922-34761922 on Build GRCh38
rs201000959 Chr.1: 34761984-34761984 on Build GRCh38
rs773395030 Chr.1: 34761843-34761843 on Build GRCh38
rs779909279 Chr.1: 34761895-34761895 on Build GRCh38
rs148656277 Chr.1: 34762024-34762024 on Build GRCh38
rs3738346 Chr.1: 34761865-34761865 on Build GRCh38
rs765716688 Chr.1: 34761988-34761988 on Build GRCh38
rs777430011 Chr.1: 34761845-34761845 on Build GRCh38
rs758544896 Chr.1: 34761878-34761878 on Build GRCh38
rs756019816 Chr.1: 34762025-34762025 on Build GRCh38
rs185327282 Chr.1: 34761933-34761933 on Build GRCh38
rs180965135 Chr.1: 34761918-34761918 on Build GRCh38
rs561519962 Chr.1: 34761851-34761851 on Build GRCh38
rs768318044 Chr.1: 34761934-34761934 on Build GRCh38
rs778129905 Chr.1: 34761868-34761868 on Build GRCh38
rs200936960 Chr.1: 34761921-34761921 on Build GRCh38
rs769791668 Chr.1: 34761949-34761949 on Build GRCh38
rs756737829 Chr.1: 34761880-34761880 on Build GRCh38
rs147456169 Chr.1: 34761899-34761899 on Build GRCh38
rs753166307 Chr.1: 34761910-34761910 on Build GRCh38
rs568452902 Chr.1: 34761952-34761952 on Build GRCh38
rs766751826 Chr.1: 34761846-34761846 on Build GRCh38
rs749997834 Chr.1: 34761989-34761989 on Build GRCh38
rs754364013 Chr.1: 34761919-34761919 on Build GRCh38
rs759786284 Chr.1: 34761857-34761857 on Build GRCh38
rs764398093 Chr.1: 34761965-34761965 on Build GRCh38
rs565868320 Chr.1: 34761947-34761947 on Build GRCh38
rs548184524 Chr.1: 34761962-34761962 on Build GRCh38
rs774534707 Chr.1: 34761907-34761907 on Build GRCh38
rs753414684 Chr.1: 34761879-34761879 on Build GRCh38
rs549139696 Chr.1: 34761829-34761829 on Build GRCh38
rs199996022 Chr.1: 34762017-34762017 on Build GRCh38
rs80142792 Chr.1: 34761881-34761881 on Build GRCh38
rs760662564 Chr.1: 34761831-34761831 on Build GRCh38
rs752939052 Chr.1: 34761862-34761862 on Build GRCh38
rs776912649 Chr.1: 34761975-34761975 on Build GRCh38
rs145404340 Chr.1: 34761953-34761953 on Build GRCh38
rs980447753 Chr.1: 34761982-34761982 on Build GRCh38
rs1033572727 Chr.1: 34761832-34761832 on Build GRCh38
rs751196580 Chr.1: 34762013-34762013 on Build GRCh38
rs775812981 Chr.1: 34761931-34761931 on Build GRCh38
rs1033213319 Chr.1: 34762001-34762001 on Build GRCh38
rs556066067 Chr.1: 34762018-34762018 on Build GRCh38
rs1010697749 Chr.1: 34762022-34762022 on Build GRCh38
rs766101103 Chr.1: 34762012-34762012 on Build GRCh38
rs777849647 Chr.1: 34762027-34762027 on Build GRCh38
rs565502726 Chr.1: 34761905-34761905 on Build GRCh38
rs771645189 Chr.1: 34761892-34761892 on Build GRCh38
rs190194468 Chr.1: 34761963-34761963 on Build GRCh38
rs1024273124 Chr.1: 34761939-34761939 on Build GRCh38

View Full Product Details