Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93692455-93692722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRA10AC1
Entrez Gene ID
118924
Gene Name
fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1
Gene Aliases
C10orf4, F26C11.1-like, FRA10A
UniGene
Hs.586650
Gene Chromosome Location
Chr.10: 93667883-93702572 on Build GRCh38
Transcripts
NM_145246.4 XM_005269515.4 XM_005269516.4 XM_011539259.2 XM_011539260.2 XM_017015656.1 XM_017015657.1 XM_017015658.1 XM_017015659.1 XM_017015660.1 XM_017015661.1 XM_017015662.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4626703 c.363C>T p.D121D Chr.10: 93692663-93692663 on Build GRCh38
3441724 c.345C>T p.F115F Chr.10: 93692681-93692681 on Build GRCh38
3721853 c.322G>A p.V108I Chr.10: 93692704-93692704 on Build GRCh38
3441722 c.345C>T p.F115F Chr.10: 93692681-93692681 on Build GRCh38
3441725 c.345C>T p.F115F Chr.10: 93692681-93692681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538983463 Chr.10: 93692518-93692518 on Build GRCh38
rs761034126 Chr.10: 93692673-93692673 on Build GRCh38
rs765938342 Chr.10: 93692662-93692662 on Build GRCh38
rs764388352 Chr.10: 93692689-93692689 on Build GRCh38
rs762703626 Chr.10: 93692651-93692651 on Build GRCh38
rs775993516 Chr.10: 93692665-93692665 on Build GRCh38
rs748036832 Chr.10: 93692637-93692637 on Build GRCh38
rs370163492 Chr.10: 93692627-93692627 on Build GRCh38
rs746133904 Chr.10: 93692604-93692604 on Build GRCh38
rs968907595 Chr.10: 93692484-93692484 on Build GRCh38
rs879206291 Chr.10: 93692498-93692498 on Build GRCh38
rs778810738 Chr.10: 93692594-93692594 on Build GRCh38
rs1000548302 Chr.10: 93692529-93692529 on Build GRCh38
rs558825722 Chr.10: 93692527-93692527 on Build GRCh38
rs776712967 Chr.10: 93692628-93692628 on Build GRCh38
rs757344367 Chr.10: 93692593-93692593 on Build GRCh38
rs929811512 Chr.10: 93692663-93692663 on Build GRCh38
rs769366235 Chr.10: 93692638-93692638 on Build GRCh38
rs73312804 Chr.10: 93692626-93692626 on Build GRCh38
rs956353597 Chr.10: 93692570-93692570 on Build GRCh38
rs1013314784 Chr.10: 93692588-93692588 on Build GRCh38
rs566162464 Chr.10: 93692536-93692536 on Build GRCh38
rs903733334 Chr.10: 93692525-93692525 on Build GRCh38
rs758517925 Chr.10: 93692619-93692619 on Build GRCh38
rs772552456 Chr.10: 93692647-93692647 on Build GRCh38
rs942697622 Chr.10: 93692603-93692603 on Build GRCh38
rs1031058942 Chr.10: 93692566-93692566 on Build GRCh38

View Full Product Details