Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77902977-77903238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDNRB
Entrez Gene ID
1910
Gene Name
endothelin receptor type B
Gene Aliases
ABCDS, ET-B, ET-BR, ETB, ETB1, ETBR, ETRB, HSCR, HSCR2, WS4A
UniGene
Hs.732046
Gene Chromosome Location
Chr.13: 77895481-77975529 on Build GRCh38
Transcripts
NM_000115.3 NM_001122659.2 NM_001201397.1 NM_003991.3 NR_103853.1

Gene Symbol
EDNRB-AS1
Entrez Gene ID
100505518
Gene Name
EDNRB antisense RNA 1
Gene Aliases
-
UniGene
Hs.742215
Gene Chromosome Location
Chr.13: 77818937-77908442 on Build GRCh38
Transcripts
NM_000115.3 NM_001122659.2 NM_001201397.1 NM_003991.3 NR_103853.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
432583 c.787A>G p.T263A Chr.13: 77903170-77903170 on Build GRCh38
4680212 c.777C>T p.P259P Chr.13: 77903180-77903180 on Build GRCh38
6139576 c.727A>G p.I243V Chr.13: 77903230-77903230 on Build GRCh38
1950562 c.1027C>T p.R343* Chr.13: 77903200-77903200 on Build GRCh38
4457371 c.1042C>T p.H348Y Chr.13: 77903185-77903185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754040927 Chr.13: 77903202-77903202 on Build GRCh38
rs140514830 Chr.13: 77903199-77903199 on Build GRCh38
rs104894390 Chr.13: 77903200-77903200 on Build GRCh38
rs375637651 Chr.13: 77903180-77903180 on Build GRCh38
rs879113182 Chr.13: 77903184-77903184 on Build GRCh38
rs77132068 Chr.13: 77903179-77903179 on Build GRCh38
rs377488350 Chr.13: 77903127-77903127 on Build GRCh38
rs977380746 Chr.13: 77903204-77903204 on Build GRCh38
rs370977355 Chr.13: 77903136-77903136 on Build GRCh38
rs899618795 Chr.13: 77903025-77903025 on Build GRCh38
rs750477425 Chr.13: 77903197-77903197 on Build GRCh38
rs1033388767 Chr.13: 77903138-77903138 on Build GRCh38
rs200431358 Chr.13: 77903182-77903182 on Build GRCh38
rs530578139 Chr.13: 77903094-77903094 on Build GRCh38
rs768394514 Chr.13: 77903114-77903114 on Build GRCh38
rs932392755 Chr.13: 77903066-77903067 on Build GRCh38
rs749117704 Chr.13: 77903113-77903113 on Build GRCh38
rs201072847 Chr.13: 77903174-77903174 on Build GRCh38
rs772585011 Chr.13: 77903131-77903131 on Build GRCh38
rs368940609 Chr.13: 77903185-77903185 on Build GRCh38
rs570185768 Chr.13: 77903147-77903147 on Build GRCh38
rs748047611 Chr.13: 77903125-77903125 on Build GRCh38
rs778871426 Chr.13: 77903124-77903124 on Build GRCh38
rs3027107 Chr.13: 77903008-77903008 on Build GRCh38
rs371057149 Chr.13: 77903196-77903196 on Build GRCh38
rs147868826 Chr.13: 77903108-77903108 on Build GRCh38
rs890940624 Chr.13: 77903089-77903089 on Build GRCh38
rs1009840735 Chr.13: 77903060-77903060 on Build GRCh38
rs892381661 Chr.13: 77903046-77903046 on Build GRCh38
rs1046252347 Chr.13: 77903100-77903100 on Build GRCh38
rs752147666 Chr.13: 77903183-77903183 on Build GRCh38
rs1051385989 Chr.13: 77903073-77903073 on Build GRCh38

View Full Product Details