Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69209722-69209949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPH
Entrez Gene ID
64946
Gene Name
centromere protein H
Gene Aliases
-
UniGene
Hs.631967
Gene Chromosome Location
Chr.5: 69189548-69210357 on Build GRCh38
Transcripts
NM_022909.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3669472 c.695C>A p.A232D Chr.5: 69209750-69209750 on Build GRCh38
6171576 c.738G>T p.M246I Chr.5: 69209793-69209793 on Build GRCh38
7314207 c.709G>T p.V237F Chr.5: 69209764-69209764 on Build GRCh38
4505558 c.696C>T p.A232A Chr.5: 69209751-69209751 on Build GRCh38
7186878 c.691C>A p.P231T Chr.5: 69209746-69209746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750302807 Chr.5: 69209847-69209847 on Build GRCh38
rs999229140 Chr.5: 69209813-69209813 on Build GRCh38
rs771076584 Chr.5: 69209774-69209774 on Build GRCh38
rs747527479 Chr.5: 69209770-69209770 on Build GRCh38
rs767304204 Chr.5: 69209821-69209821 on Build GRCh38
rs765537561 Chr.5: 69209793-69209793 on Build GRCh38
rs757451437 Chr.5: 69209888-69209888 on Build GRCh38
rs372930981 Chr.5: 69209882-69209882 on Build GRCh38
rs1003305735 Chr.5: 69209854-69209854 on Build GRCh38
rs375648256 Chr.5: 69209785-69209785 on Build GRCh38
rs902654472 Chr.5: 69209852-69209852 on Build GRCh38
rs764466772 Chr.5: 69209876-69209876 on Build GRCh38
rs765798184 Chr.5: 69209851-69209851 on Build GRCh38
rs374838407 Chr.5: 69209871-69209874 on Build GRCh38
rs775595443 Chr.5: 69209803-69209803 on Build GRCh38
rs761555014 Chr.5: 69209817-69209817 on Build GRCh38
rs548488447 Chr.5: 69209883-69209883 on Build GRCh38
rs756098489 Chr.5: 69209850-69209850 on Build GRCh38
rs978193540 Chr.5: 69209867-69209867 on Build GRCh38
rs1042535015 Chr.5: 69209837-69209837 on Build GRCh38
rs776830111 Chr.5: 69209776-69209776 on Build GRCh38
rs367681440 Chr.5: 69209909-69209909 on Build GRCh38

View Full Product Details