Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100874264-100874501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL2
Entrez Gene ID
2135
Gene Name
exostosin like glycosyltransferase 2
Gene Aliases
EXTR2
UniGene
Hs.357637
Gene Chromosome Location
Chr.1: 100872372-100895411 on Build GRCh38
Transcripts
NM_001033025.2 NM_001261440.1 NM_001261441.1 NM_001261442.1 NM_001439.3 NR_048570.1 XM_005270621.1 XM_011540994.2 XM_011540995.2 XM_011540996.2 XM_017000651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3470329 c.573T>C p.S191S Chr.1: 100874362-100874362 on Build GRCh38
6019512 c.560C>G p.S187* Chr.1: 100874375-100874375 on Build GRCh38
6019513 c.584C>G p.S195* Chr.1: 100874375-100874375 on Build GRCh38
5735558 c.505C>T p.Q169* Chr.1: 100874430-100874430 on Build GRCh38
5735559 c.529C>T p.Q177* Chr.1: 100874430-100874430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926689168 Chr.1: 100874360-100874360 on Build GRCh38
rs752104887 Chr.1: 100874422-100874422 on Build GRCh38
rs374572592 Chr.1: 100874389-100874389 on Build GRCh38
rs779939389 Chr.1: 100874391-100874391 on Build GRCh38
rs775788632 Chr.1: 100874452-100874452 on Build GRCh38
rs759882915 Chr.1: 100874334-100874334 on Build GRCh38
rs968226491 Chr.1: 100874335-100874335 on Build GRCh38
rs147248391 Chr.1: 100874315-100874315 on Build GRCh38
rs80119413 Chr.1: 100874464-100874464 on Build GRCh38
rs770304647 Chr.1: 100874438-100874438 on Build GRCh38
rs1031376839 Chr.1: 100874301-100874301 on Build GRCh38
rs377016982 Chr.1: 100874359-100874359 on Build GRCh38
rs371011616 Chr.1: 100874384-100874384 on Build GRCh38
rs777121235 Chr.1: 100874423-100874423 on Build GRCh38
rs549748136 Chr.1: 100874370-100874370 on Build GRCh38
rs777414628 Chr.1: 100874467-100874467 on Build GRCh38
rs756123522 Chr.1: 100874390-100874390 on Build GRCh38
rs184097400 Chr.1: 100874421-100874421 on Build GRCh38
rs141823466 Chr.1: 100874307-100874307 on Build GRCh38
rs746310275 Chr.1: 100874435-100874435 on Build GRCh38
rs769675035 Chr.1: 100874311-100874312 on Build GRCh38
rs749120908 Chr.1: 100874403-100874403 on Build GRCh38
rs758610643 Chr.1: 100874412-100874412 on Build GRCh38
rs982314186 Chr.1: 100874349-100874349 on Build GRCh38
rs751049020 Chr.1: 100874372-100874372 on Build GRCh38
rs765659323 Chr.1: 100874356-100874356 on Build GRCh38
rs767472992 Chr.1: 100874388-100874388 on Build GRCh38
rs1041455229 Chr.1: 100874377-100874377 on Build GRCh38
rs768327965 Chr.1: 100874469-100874469 on Build GRCh38

View Full Product Details