Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34208062-34208331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf94
Entrez Gene ID
84970
Gene Name
chromosome 1 open reading frame 94
Gene Aliases
-
UniGene
Hs.194610
Gene Chromosome Location
Chr.1: 34166883-34219131 on Build GRCh38
Transcripts
NM_001134734.1 NM_032884.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5010520 c.1478C>G p.P493R Chr.1: 34208188-34208188 on Build GRCh38
1342059 c.1469C>T p.A490V Chr.1: 34208179-34208179 on Build GRCh38
5527674 c.903G>A p.R301R Chr.1: 34208183-34208183 on Build GRCh38
1342061 c.1490_1491insT p.L498fs*>102 Chr.1: 34208200-34208201 on Build GRCh38
1687429 c.878G>A p.G293D Chr.1: 34208158-34208158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746203826 Chr.1: 34208245-34208245 on Build GRCh38
rs372631054 Chr.1: 34208152-34208152 on Build GRCh38
rs748206649 Chr.1: 34208129-34208129 on Build GRCh38
rs776039610 Chr.1: 34208276-34208282 on Build GRCh38
rs769388080 Chr.1: 34208274-34208274 on Build GRCh38
rs891278168 Chr.1: 34208097-34208097 on Build GRCh38
rs746560436 Chr.1: 34208266-34208266 on Build GRCh38
rs749731038 Chr.1: 34208135-34208135 on Build GRCh38
rs761661462 Chr.1: 34208191-34208191 on Build GRCh38
rs774895568 Chr.1: 34208167-34208167 on Build GRCh38
rs776304539 Chr.1: 34208272-34208272 on Build GRCh38
rs758286207 Chr.1: 34208219-34208219 on Build GRCh38
rs750107449 Chr.1: 34208193-34208193 on Build GRCh38
rs41266413 Chr.1: 34208100-34208100 on Build GRCh38
rs756203172 Chr.1: 34208235-34208235 on Build GRCh38
rs752720598 Chr.1: 34208228-34208228 on Build GRCh38
rs754085272 Chr.1: 34208241-34208241 on Build GRCh38
rs917828178 Chr.1: 34208151-34208151 on Build GRCh38
rs866875499 Chr.1: 34208223-34208223 on Build GRCh38
rs761432043 Chr.1: 34208273-34208273 on Build GRCh38
rs564492135 Chr.1: 34208210-34208210 on Build GRCh38
rs750322569 Chr.1: 34208110-34208110 on Build GRCh38
rs781109634 Chr.1: 34208122-34208122 on Build GRCh38
rs762738791 Chr.1: 34208285-34208285 on Build GRCh38
rs529846578 Chr.1: 34208291-34208291 on Build GRCh38
rs1007094373 Chr.1: 34208099-34208099 on Build GRCh38
rs115970539 Chr.1: 34208180-34208180 on Build GRCh38
rs757924582 Chr.1: 34208216-34208216 on Build GRCh38
rs778210618 Chr.1: 34208237-34208237 on Build GRCh38
rs114229205 Chr.1: 34208244-34208244 on Build GRCh38
rs79398834 Chr.1: 34208257-34208257 on Build GRCh38
rs766297459 Chr.1: 34208224-34208224 on Build GRCh38
rs924071872 Chr.1: 34208117-34208117 on Build GRCh38
rs755125096 Chr.1: 34208114-34208114 on Build GRCh38
rs764984501 Chr.1: 34208209-34208209 on Build GRCh38
rs201753293 Chr.1: 34208158-34208158 on Build GRCh38
rs568183080 Chr.1: 34208268-34208268 on Build GRCh38
rs866186850 Chr.1: 34208125-34208125 on Build GRCh38
rs772908085 Chr.1: 34208280-34208280 on Build GRCh38
rs923977955 Chr.1: 34208208-34208208 on Build GRCh38
rs768148966 Chr.1: 34208267-34208267 on Build GRCh38
rs371631981 Chr.1: 34208145-34208145 on Build GRCh38
rs960818941 Chr.1: 34208311-34208311 on Build GRCh38
rs776249666 Chr.1: 34208182-34208182 on Build GRCh38
rs760165855 Chr.1: 34208179-34208179 on Build GRCh38
rs756432508 Chr.1: 34208130-34208130 on Build GRCh38
rs772462855 Chr.1: 34208252-34208252 on Build GRCh38

View Full Product Details