Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108518688-108518933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FICD
Entrez Gene ID
11153
Gene Name
FIC domain containing
Gene Aliases
HIP13, HYPE, UNQ3041
UniGene
Hs.661891
Gene Chromosome Location
Chr.12: 108515274-108519603 on Build GRCh38
Transcripts
NM_007076.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5051303 c.820A>G p.I274V Chr.12: 108518918-108518918 on Build GRCh38
161071 c.611A>C p.K204T Chr.12: 108518709-108518709 on Build GRCh38
1605523 c.630G>T p.K210N Chr.12: 108518728-108518728 on Build GRCh38
5601500 c.755C>A p.A252D Chr.12: 108518853-108518853 on Build GRCh38
5636386 c.651G>T p.R217S Chr.12: 108518749-108518749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034220285 Chr.12: 108518817-108518817 on Build GRCh38
rs756645337 Chr.12: 108518798-108518798 on Build GRCh38
rs767630290 Chr.12: 108518855-108518855 on Build GRCh38
rs763105184 Chr.12: 108518768-108518768 on Build GRCh38
rs745535049 Chr.12: 108518881-108518881 on Build GRCh38
rs559793492 Chr.12: 108518800-108518800 on Build GRCh38
rs761628825 Chr.12: 108518850-108518850 on Build GRCh38
rs748131616 Chr.12: 108518897-108518897 on Build GRCh38
rs770170665 Chr.12: 108518729-108518729 on Build GRCh38
rs747099327 Chr.12: 108518858-108518858 on Build GRCh38
rs779462728 Chr.12: 108518873-108518873 on Build GRCh38
rs771220652 Chr.12: 108518759-108518760 on Build GRCh38
rs764699967 Chr.12: 108518860-108518860 on Build GRCh38
rs781177626 Chr.12: 108518795-108518795 on Build GRCh38
rs757676830 Chr.12: 108518862-108518862 on Build GRCh38
rs371170438 Chr.12: 108518861-108518861 on Build GRCh38
rs866522242 Chr.12: 108518822-108518822 on Build GRCh38
rs151147125 Chr.12: 108518744-108518744 on Build GRCh38
rs35990156 Chr.12: 108518766-108518766 on Build GRCh38
rs376371078 Chr.12: 108518859-108518859 on Build GRCh38
rs899105696 Chr.12: 108518812-108518812 on Build GRCh38
rs142218882 Chr.12: 108518821-108518821 on Build GRCh38
rs146316463 Chr.12: 108518844-108518844 on Build GRCh38
rs140164827 Chr.12: 108518797-108518797 on Build GRCh38
rs372551468 Chr.12: 108518846-108518846 on Build GRCh38
rs1000962338 Chr.12: 108518781-108518782 on Build GRCh38
rs751598998 Chr.12: 108518788-108518788 on Build GRCh38
rs776345995 Chr.12: 108518847-108518847 on Build GRCh38
rs552820677 Chr.12: 108518852-108518852 on Build GRCh38
rs545390876 Chr.12: 108518892-108518892 on Build GRCh38
rs368813727 Chr.12: 108518791-108518791 on Build GRCh38
rs766593011 Chr.12: 108518770-108518770 on Build GRCh38
rs778321019 Chr.12: 108518738-108518738 on Build GRCh38
rs190616191 Chr.12: 108518749-108518749 on Build GRCh38
rs759118075 Chr.12: 108518746-108518746 on Build GRCh38
rs770637083 Chr.12: 108518819-108518819 on Build GRCh38
rs779559277 Chr.12: 108518891-108518891 on Build GRCh38
rs774932592 Chr.12: 108518830-108518830 on Build GRCh38
rs147612703 Chr.12: 108518815-108518815 on Build GRCh38
rs745463913 Chr.12: 108518756-108518756 on Build GRCh38
rs778735949 Chr.12: 108518820-108518820 on Build GRCh38
rs372339284 Chr.12: 108518767-108518767 on Build GRCh38
rs199846789 Chr.12: 108518851-108518851 on Build GRCh38
rs769545515 Chr.12: 108518898-108518898 on Build GRCh38
rs372982792 Chr.12: 108518750-108518750 on Build GRCh38
rs773935794 Chr.12: 108518745-108518745 on Build GRCh38
rs771584835 Chr.12: 108518828-108518828 on Build GRCh38
rs748941881 Chr.12: 108518739-108518739 on Build GRCh38
rs150326458 Chr.12: 108518809-108518809 on Build GRCh38
rs748412077 Chr.12: 108518722-108518722 on Build GRCh38
rs776573134 Chr.12: 108518896-108518896 on Build GRCh38
rs772602329 Chr.12: 108518839-108518839 on Build GRCh38
rs759715090 Chr.12: 108518854-108518854 on Build GRCh38
rs758048881 Chr.12: 108518890-108518890 on Build GRCh38

View Full Product Details