Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 34028713-34028954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF3
Entrez Gene ID
114899
Gene Name
C1q and tumor necrosis factor related protein 3
Gene Aliases
C1ATNF3, CORCS, CORS, CORS-26, CORS26, CTRP3
UniGene
Hs.171929
Gene Chromosome Location
Chr.5: 34017858-34043266 on Build GRCh38
Transcripts
NM_030945.3 NM_181435.5 NR_037951.1

Gene Symbol
C1QTNF3-AMACR
Entrez Gene ID
100534612
Gene Name
C1QTNF3-AMACR readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.171929
Gene Chromosome Location
Chr.5: 33986986-34124528 on Build GRCh38
Transcripts
NM_030945.3 NM_181435.5 NR_037951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3702800 c.571-2A>C p.? Chr.5: 34028885-34028885 on Build GRCh38
6171072 c.630C>T p.F210F Chr.5: 34028824-34028824 on Build GRCh38
5356423 c.571-10C>A p.? Chr.5: 34028893-34028893 on Build GRCh38
7358342 c.646A>T p.N216Y Chr.5: 34028808-34028808 on Build GRCh38
213785 c.612G>T p.Q204H Chr.5: 34028842-34028842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755141201 Chr.5: 34028805-34028805 on Build GRCh38
rs984445039 Chr.5: 34028841-34028841 on Build GRCh38
rs762252653 Chr.5: 34028762-34028762 on Build GRCh38
rs766726884 Chr.5: 34028902-34028902 on Build GRCh38
rs749014950 Chr.5: 34028820-34028820 on Build GRCh38
rs745473321 Chr.5: 34028842-34028842 on Build GRCh38
rs774917120 Chr.5: 34028857-34028857 on Build GRCh38
rs778458732 Chr.5: 34028839-34028839 on Build GRCh38
rs756115401 Chr.5: 34028816-34028816 on Build GRCh38
rs767770851 Chr.5: 34028917-34028917 on Build GRCh38
rs751701557 Chr.5: 34028804-34028804 on Build GRCh38
rs144863962 Chr.5: 34028921-34028921 on Build GRCh38
rs765593973 Chr.5: 34028798-34028798 on Build GRCh38
rs767658386 Chr.5: 34028808-34028808 on Build GRCh38
rs748724274 Chr.5: 34028861-34028861 on Build GRCh38
rs777768121 Chr.5: 34028818-34028818 on Build GRCh38
rs371598034 Chr.5: 34028803-34028803 on Build GRCh38
rs375895254 Chr.5: 34028890-34028890 on Build GRCh38
rs763298981 Chr.5: 34028802-34028802 on Build GRCh38
rs1031149128 Chr.5: 34028889-34028889 on Build GRCh38
rs199873714 Chr.5: 34028867-34028867 on Build GRCh38
rs774459574 Chr.5: 34028905-34028905 on Build GRCh38
rs752684956 Chr.5: 34028815-34028815 on Build GRCh38
rs200967666 Chr.5: 34028827-34028827 on Build GRCh38
rs777143468 Chr.5: 34028749-34028749 on Build GRCh38
rs771581816 Chr.5: 34028854-34028854 on Build GRCh38
rs773449408 Chr.5: 34028800-34028800 on Build GRCh38
rs759776301 Chr.5: 34028908-34028908 on Build GRCh38

View Full Product Details