Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53210741-53211014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPT2
Entrez Gene ID
1376
Gene Name
carnitine palmitoyltransferase 2
Gene Aliases
CPT1, CPTASE, IIAE4
UniGene
Hs.713535
Gene Chromosome Location
Chr.1: 53196429-53214197 on Build GRCh38
Transcripts
NM_000098.2 XM_005270484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6063693 c.1251C>T p.F417F Chr.1: 53210925-53210925 on Build GRCh38
3934781 c.1184C>A p.P395H Chr.1: 53210858-53210858 on Build GRCh38
4577444 c.1189G>A p.V397I Chr.1: 53210863-53210863 on Build GRCh38
6009286 c.1074C>G p.L358L Chr.1: 53210748-53210748 on Build GRCh38
3751293 c.1102G>A p.V368I Chr.1: 53210776-53210776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757953266 Chr.1: 53210948-53210948 on Build GRCh38
rs773474888 Chr.1: 53210883-53210883 on Build GRCh38
rs576822710 Chr.1: 53210925-53210925 on Build GRCh38
rs767826599 Chr.1: 53210862-53210862 on Build GRCh38
rs777506825 Chr.1: 53210940-53210941 on Build GRCh38
rs865856450 Chr.1: 53210978-53210978 on Build GRCh38
rs760514258 Chr.1: 53210811-53210811 on Build GRCh38
rs397509431 Chr.1: 53210912-53210913 on Build GRCh38
rs376445618 Chr.1: 53210952-53210952 on Build GRCh38
rs758408111 Chr.1: 53210832-53210832 on Build GRCh38
rs515726176 Chr.1: 53210819-53210819 on Build GRCh38
rs771486286 Chr.1: 53210795-53210795 on Build GRCh38
rs867886479 Chr.1: 53210926-53210926 on Build GRCh38
rs775246158 Chr.1: 53210984-53210984 on Build GRCh38
rs751719701 Chr.1: 53210845-53210845 on Build GRCh38
rs746567260 Chr.1: 53210902-53210902 on Build GRCh38
rs144875040 Chr.1: 53210812-53210812 on Build GRCh38
rs368989271 Chr.1: 53210927-53210927 on Build GRCh38
rs373420308 Chr.1: 53210933-53210933 on Build GRCh38
rs781398541 Chr.1: 53210766-53210766 on Build GRCh38
rs1799821 Chr.1: 53210776-53210776 on Build GRCh38
rs772572668 Chr.1: 53210899-53210899 on Build GRCh38
rs146670074 Chr.1: 53210876-53210876 on Build GRCh38
rs544400632 Chr.1: 53210910-53210910 on Build GRCh38
rs757979320 Chr.1: 53210794-53210795 on Build GRCh38
rs772843417 Chr.1: 53210798-53210798 on Build GRCh38
rs201745292 Chr.1: 53210863-53210863 on Build GRCh38
rs761347926 Chr.1: 53210790-53210790 on Build GRCh38
rs996410960 Chr.1: 53210904-53210904 on Build GRCh38
rs187044944 Chr.1: 53210958-53210958 on Build GRCh38
rs745804213 Chr.1: 53210971-53210971 on Build GRCh38
rs374201361 Chr.1: 53210986-53210986 on Build GRCh38
rs746186600 Chr.1: 53210869-53210869 on Build GRCh38
rs938668766 Chr.1: 53210774-53210774 on Build GRCh38
rs747794860 Chr.1: 53210895-53210895 on Build GRCh38
rs763073192 Chr.1: 53210916-53210916 on Build GRCh38
rs776488578 Chr.1: 53210964-53210964 on Build GRCh38
rs377616144 Chr.1: 53210987-53210987 on Build GRCh38
rs369847007 Chr.1: 53210817-53210817 on Build GRCh38
rs375957043 Chr.1: 53210906-53210906 on Build GRCh38
rs748181550 Chr.1: 53210836-53210836 on Build GRCh38
rs752932153 Chr.1: 53210829-53210829 on Build GRCh38
rs575447822 Chr.1: 53210908-53210908 on Build GRCh38
rs771811692 Chr.1: 53210897-53210897 on Build GRCh38
rs777711968 Chr.1: 53210842-53210842 on Build GRCh38
rs752373512 Chr.1: 53210895-53210896 on Build GRCh38
rs767852112 Chr.1: 53210929-53210929 on Build GRCh38
rs769781310 Chr.1: 53210976-53210976 on Build GRCh38
rs757683330 Chr.1: 53210849-53210849 on Build GRCh38
rs398123153 Chr.1: 53210913-53210914 on Build GRCh38
rs201008705 Chr.1: 53210921-53210921 on Build GRCh38
rs372102993 Chr.1: 53210903-53210903 on Build GRCh38
rs74315295 Chr.1: 53210822-53210822 on Build GRCh38
rs749714778 Chr.1: 53210937-53210937 on Build GRCh38
rs112914907 Chr.1: 53210907-53210907 on Build GRCh38
rs774168365 Chr.1: 53210784-53210784 on Build GRCh38
rs746284903 Chr.1: 53210771-53210771 on Build GRCh38
rs1024753743 Chr.1: 53210992-53210992 on Build GRCh38
rs778639977 Chr.1: 53210888-53210888 on Build GRCh38
rs770491351 Chr.1: 53210962-53210962 on Build GRCh38

View Full Product Details