Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180997106-180997375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL3
Entrez Gene ID
10917
Gene Name
butyrophilin like 3
Gene Aliases
BTN9.1, BTNLR
UniGene
Hs.225949
Gene Chromosome Location
Chr.5: 180988845-181006727 on Build GRCh38
Transcripts
NM_197975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4394701 c.537C>T p.S179S Chr.5: 180997352-180997352 on Build GRCh38
3744816 c.475T>A p.W159R Chr.5: 180997290-180997290 on Build GRCh38
4496689 c.480C>T p.F160F Chr.5: 180997295-180997295 on Build GRCh38
5558561 c.410T>C p.L137P Chr.5: 180997225-180997225 on Build GRCh38
737431 c.441C>A p.D147E Chr.5: 180997256-180997256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757711506 Chr.5: 180997242-180997242 on Build GRCh38
rs368730382 Chr.5: 180997218-180997218 on Build GRCh38
rs1042117903 Chr.5: 180997152-180997152 on Build GRCh38
rs200761034 Chr.5: 180997309-180997309 on Build GRCh38
rs776456516 Chr.5: 180997183-180997183 on Build GRCh38
rs367824060 Chr.5: 180997280-180997280 on Build GRCh38
rs949028125 Chr.5: 180997198-180997198 on Build GRCh38
rs768599711 Chr.5: 180997353-180997353 on Build GRCh38
rs764823723 Chr.5: 180997191-180997191 on Build GRCh38
rs916142366 Chr.5: 180997161-180997161 on Build GRCh38
rs990377144 Chr.5: 180997140-180997140 on Build GRCh38
rs375050350 Chr.5: 180997181-180997181 on Build GRCh38
rs371364543 Chr.5: 180997223-180997223 on Build GRCh38
rs761545927 Chr.5: 180997185-180997185 on Build GRCh38
rs867218663 Chr.5: 180997321-180997321 on Build GRCh38
rs77077795 Chr.5: 180997241-180997241 on Build GRCh38
rs749486740 Chr.5: 180997245-180997246 on Build GRCh38
rs747099872 Chr.5: 180997336-180997336 on Build GRCh38
rs982000260 Chr.5: 180997205-180997205 on Build GRCh38
rs202123434 Chr.5: 180997229-180997229 on Build GRCh38
rs750088201 Chr.5: 180997206-180997206 on Build GRCh38
rs779214012 Chr.5: 180997256-180997256 on Build GRCh38
rs930979416 Chr.5: 180997291-180997291 on Build GRCh38
rs909023655 Chr.5: 180997324-180997324 on Build GRCh38
rs867199883 Chr.5: 180997244-180997245 on Build GRCh38

View Full Product Details