Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51934805-51935058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP7B
Entrez Gene ID
540
Gene Name
ATPase copper transporting beta
Gene Aliases
PWD, WC1, WD, WND
UniGene
Hs.492280
Gene Chromosome Location
Chr.13: 51932669-52012130 on Build GRCh38
Transcripts
NM_000053.3 NM_001005918.2 NM_001243182.1 XM_005266423.2 XM_005266424.4 XM_005266427.2 XM_005266428.1 XM_005266430.4 XM_005266431.3 XM_005266432.2 XM_006719837.3 XM_006719838.1 XM_006719839.1 XM_011535117.2 XM_011535118.1 XM_011535119.1 XM_011535122.2 XM_017020627.1 XM_017020628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5913481 c.4295C>T p.S1432F Chr.13: 51934859-51934859 on Build GRCh38
6335053 c.4331C>A p.A1444E Chr.13: 51934823-51934823 on Build GRCh38
279073 c.4307A>G p.D1436G Chr.13: 51934847-51934847 on Build GRCh38
1740005 c.4187C>G p.T1396R Chr.13: 51934967-51934967 on Build GRCh38
6404390 c.4201A>G p.S1401G Chr.13: 51934953-51934953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767721448 Chr.13: 51934939-51934939 on Build GRCh38
rs768984875 Chr.13: 51935016-51935016 on Build GRCh38
rs73202048 Chr.13: 51934843-51934843 on Build GRCh38
rs960533033 Chr.13: 51935003-51935003 on Build GRCh38
rs374693153 Chr.13: 51934913-51934913 on Build GRCh38
rs769672624 Chr.13: 51934922-51934922 on Build GRCh38
rs762078360 Chr.13: 51934929-51934929 on Build GRCh38
rs764892434 Chr.13: 51934869-51934869 on Build GRCh38
rs778639218 Chr.13: 51934846-51934846 on Build GRCh38
rs755388647 Chr.13: 51934886-51934886 on Build GRCh38
rs766316953 Chr.13: 51934956-51934956 on Build GRCh38
rs761990581 Chr.13: 51934872-51934872 on Build GRCh38
rs899484304 Chr.13: 51934866-51934866 on Build GRCh38
rs757351029 Chr.13: 51934833-51934833 on Build GRCh38
rs754397173 Chr.13: 51934961-51934961 on Build GRCh38
rs775242151 Chr.13: 51934926-51934926 on Build GRCh38
rs116091486 Chr.13: 51934852-51934852 on Build GRCh38
rs60986317 Chr.13: 51934853-51934853 on Build GRCh38
rs199821965 Chr.13: 51934979-51934979 on Build GRCh38
rs764383437 Chr.13: 51934827-51934827 on Build GRCh38
rs755406059 Chr.13: 51934966-51934966 on Build GRCh38
rs375692175 Chr.13: 51934859-51934859 on Build GRCh38
rs746525895 Chr.13: 51934911-51934911 on Build GRCh38
rs181250704 Chr.13: 51935019-51935019 on Build GRCh38
rs1041361346 Chr.13: 51934973-51934973 on Build GRCh38
rs770900174 Chr.13: 51934855-51934855 on Build GRCh38
rs768833241 Chr.13: 51934845-51934845 on Build GRCh38
rs747816425 Chr.13: 51934867-51934867 on Build GRCh38
rs559705781 Chr.13: 51934923-51934923 on Build GRCh38
rs770290708 Chr.13: 51934912-51934912 on Build GRCh38
rs1057517191 Chr.13: 51934912-51934912 on Build GRCh38
rs549611809 Chr.13: 51934877-51934877 on Build GRCh38
rs763155011 Chr.13: 51934825-51934825 on Build GRCh38
rs760766549 Chr.13: 51935033-51935033 on Build GRCh38
rs773440614 Chr.13: 51935025-51935025 on Build GRCh38
rs543334965 Chr.13: 51934836-51934836 on Build GRCh38
rs148081616 Chr.13: 51934848-51934848 on Build GRCh38
rs779103007 Chr.13: 51934891-51934891 on Build GRCh38
rs780613139 Chr.13: 51934882-51934882 on Build GRCh38
rs777494364 Chr.13: 51934908-51934908 on Build GRCh38
rs372435824 Chr.13: 51934883-51934883 on Build GRCh38
rs939136907 Chr.13: 51934840-51934840 on Build GRCh38
rs546721020 Chr.13: 51934903-51934903 on Build GRCh38
rs201483366 Chr.13: 51934835-51934835 on Build GRCh38
rs756826065 Chr.13: 51934990-51934990 on Build GRCh38
rs759353316 Chr.13: 51934917-51934917 on Build GRCh38
rs368382723 Chr.13: 51934881-51934881 on Build GRCh38
rs371458882 Chr.13: 51934991-51934991 on Build GRCh38
rs572639840 Chr.13: 51934947-51934947 on Build GRCh38
rs780338961 Chr.13: 51934838-51934838 on Build GRCh38
rs957678118 Chr.13: 51934948-51934948 on Build GRCh38
rs745866259 Chr.13: 51934992-51934992 on Build GRCh38
rs886041336 Chr.13: 51934959-51934959 on Build GRCh38
rs746611866 Chr.13: 51934987-51934987 on Build GRCh38
rs528603867 Chr.13: 51934967-51934967 on Build GRCh38
rs587783320 Chr.13: 51934933-51934933 on Build GRCh38
rs771736533 Chr.13: 51934849-51934849 on Build GRCh38
rs769604207 Chr.13: 51935006-51935006 on Build GRCh38
rs751677241 Chr.13: 51934830-51934830 on Build GRCh38
rs767633246 Chr.13: 51934875-51934875 on Build GRCh38
rs998939884 Chr.13: 51934927-51934927 on Build GRCh38
rs775330670 Chr.13: 51935011-51935011 on Build GRCh38
rs532864705 Chr.13: 51934893-51934893 on Build GRCh38
rs908367214 Chr.13: 51935024-51935024 on Build GRCh38
rs189601972 Chr.13: 51934941-51934941 on Build GRCh38
rs776367340 Chr.13: 51934858-51934858 on Build GRCh38
rs751811446 Chr.13: 51934870-51934870 on Build GRCh38
rs777585988 Chr.13: 51934980-51934980 on Build GRCh38
rs774744497 Chr.13: 51934829-51934829 on Build GRCh38
rs368801566 Chr.13: 51934969-51934969 on Build GRCh38

View Full Product Details