Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1466742-1466995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPCAT1
Entrez Gene ID
79888
Gene Name
lysophosphatidylcholine acyltransferase 1
Gene Aliases
AGPAT10, AGPAT9, AYTL2, PFAAP3, lpcat
UniGene
Hs.368853
Gene Chromosome Location
Chr.5: 1461427-1523961 on Build GRCh38
Transcripts
NM_024830.3 XM_005248373.3 XM_011514132.1 XM_011514134.1 XM_017009879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3827336 c.1351G>T p.V451L Chr.5: 1466818-1466818 on Build GRCh38
3661578 c.1301G>A p.G434D Chr.5: 1466868-1466868 on Build GRCh38
6702320 c.1343C>T p.A448V Chr.5: 1466826-1466826 on Build GRCh38
6702317 c.1408A>G p.K470E Chr.5: 1466761-1466761 on Build GRCh38
5811106 c.1372G>T p.D458Y Chr.5: 1466797-1466797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009491999 Chr.5: 1466955-1466955 on Build GRCh38
rs535425512 Chr.5: 1466869-1466869 on Build GRCh38
rs766984997 Chr.5: 1466798-1466798 on Build GRCh38
rs1044110820 Chr.5: 1466952-1466952 on Build GRCh38
rs201936413 Chr.5: 1466905-1466905 on Build GRCh38
rs369461212 Chr.5: 1466898-1466898 on Build GRCh38
rs781199821 Chr.5: 1466812-1466812 on Build GRCh38
rs765629260 Chr.5: 1466803-1466803 on Build GRCh38
rs770874761 Chr.5: 1466792-1466792 on Build GRCh38
rs955924676 Chr.5: 1466849-1466849 on Build GRCh38
rs148166675 Chr.5: 1466794-1466794 on Build GRCh38
rs2277006 Chr.5: 1466804-1466804 on Build GRCh38
rs766712143 Chr.5: 1466888-1466888 on Build GRCh38
rs745749952 Chr.5: 1466936-1466936 on Build GRCh38
rs748381368 Chr.5: 1466783-1466783 on Build GRCh38
rs35452723 Chr.5: 1466889-1466889 on Build GRCh38
rs7718422 Chr.5: 1466926-1466926 on Build GRCh38
rs543894552 Chr.5: 1466789-1466789 on Build GRCh38
rs776889940 Chr.5: 1466778-1466778 on Build GRCh38
rs769575480 Chr.5: 1466784-1466784 on Build GRCh38
rs749557044 Chr.5: 1466914-1466914 on Build GRCh38
rs527556981 Chr.5: 1466929-1466929 on Build GRCh38
rs777938530 Chr.5: 1466902-1466902 on Build GRCh38
rs1032005944 Chr.5: 1466824-1466824 on Build GRCh38
rs760344160 Chr.5: 1466873-1466873 on Build GRCh38
rs146171733 Chr.5: 1466921-1466921 on Build GRCh38
rs778638943 Chr.5: 1466928-1466928 on Build GRCh38
rs143106036 Chr.5: 1466868-1466868 on Build GRCh38
rs771915409 Chr.5: 1466942-1466942 on Build GRCh38
rs376760316 Chr.5: 1466791-1466791 on Build GRCh38
rs752799102 Chr.5: 1466897-1466897 on Build GRCh38
rs749109602 Chr.5: 1466839-1466839 on Build GRCh38
rs747144587 Chr.5: 1466768-1466768 on Build GRCh38
rs755076181 Chr.5: 1466807-1466807 on Build GRCh38
rs1044903820 Chr.5: 1466850-1466850 on Build GRCh38
rs895963378 Chr.5: 1466946-1466946 on Build GRCh38
rs1051972897 Chr.5: 1466971-1466971 on Build GRCh38
rs1012632730 Chr.5: 1466949-1466949 on Build GRCh38
rs748337774 Chr.5: 1466816-1466816 on Build GRCh38
rs904218966 Chr.5: 1466967-1466967 on Build GRCh38
rs545604406 Chr.5: 1466948-1466948 on Build GRCh38
rs942008585 Chr.5: 1466884-1466884 on Build GRCh38
rs375037785 Chr.5: 1466855-1466855 on Build GRCh38
rs760470508 Chr.5: 1466801-1466801 on Build GRCh38
rs752381395 Chr.5: 1466800-1466800 on Build GRCh38
rs954777354 Chr.5: 1466870-1466870 on Build GRCh38
rs774319281 Chr.5: 1466861-1466861 on Build GRCh38
rs780580539 Chr.5: 1466806-1466806 on Build GRCh38
rs7714454 Chr.5: 1466919-1466919 on Build GRCh38
rs748135748 Chr.5: 1466890-1466890 on Build GRCh38
rs372740293 Chr.5: 1466885-1466885 on Build GRCh38
rs759051643 Chr.5: 1466795-1466795 on Build GRCh38
rs986179103 Chr.5: 1466872-1466872 on Build GRCh38
rs768659423 Chr.5: 1466776-1466776 on Build GRCh38
rs113155822 Chr.5: 1466817-1466817 on Build GRCh38
rs753005558 Chr.5: 1466768-1466770 on Build GRCh38
rs770019660 Chr.5: 1466828-1466828 on Build GRCh38
rs763872166 Chr.5: 1466879-1466879 on Build GRCh38

View Full Product Details