Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 42787035-42787304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMS1
Entrez Gene ID
9790
Gene Name
BMS1, ribosome biogenesis factor
Gene Aliases
ACC, BMS1L
UniGene
Hs.10848
Gene Chromosome Location
Chr.10: 42782506-42834937 on Build GRCh38
Transcripts
NM_014753.3 XM_005271846.3 XM_005271848.3 XM_005271849.3 XM_011540402.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
918084 c.376C>T p.R126C Chr.10: 42787176-42787176 on Build GRCh38
259169 c.383T>G p.L128R Chr.10: 42787183-42787183 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs916334894 Chr.10: 42787206-42787206 on Build GRCh38
rs756531858 Chr.10: 42787184-42787184 on Build GRCh38
rs769777591 Chr.10: 42787261-42787261 on Build GRCh38
rs780540295 Chr.10: 42787174-42787175 on Build GRCh38
rs771767050 Chr.10: 42787227-42787227 on Build GRCh38
rs750730619 Chr.10: 42787171-42787171 on Build GRCh38
rs774809970 Chr.10: 42787236-42787236 on Build GRCh38
rs778536019 Chr.10: 42787188-42787188 on Build GRCh38
rs565731481 Chr.10: 42787242-42787242 on Build GRCh38
rs191919925 Chr.10: 42787114-42787114 on Build GRCh38
rs972624226 Chr.10: 42787120-42787120 on Build GRCh38
rs763036596 Chr.10: 42787267-42787267 on Build GRCh38
rs781697087 Chr.10: 42787178-42787178 on Build GRCh38
rs761740448 Chr.10: 42787164-42787164 on Build GRCh38
rs773156244 Chr.10: 42787262-42787262 on Build GRCh38
rs765532200 Chr.10: 42787165-42787165 on Build GRCh38
rs766655500 Chr.10: 42787274-42787274 on Build GRCh38
rs755400315 Chr.10: 42787177-42787177 on Build GRCh38
rs533191255 Chr.10: 42787254-42787254 on Build GRCh38
rs551370809 Chr.10: 42787276-42787276 on Build GRCh38
rs561943045 Chr.10: 42787118-42787118 on Build GRCh38
rs11239772 Chr.10: 42787080-42787080 on Build GRCh38
rs564212508 Chr.10: 42787084-42787084 on Build GRCh38
rs547314695 Chr.10: 42787202-42787202 on Build GRCh38
rs776525557 Chr.10: 42787147-42787147 on Build GRCh38
rs529159115 Chr.10: 42787176-42787176 on Build GRCh38
rs766314874 Chr.10: 42787174-42787174 on Build GRCh38
rs531447303 Chr.10: 42787102-42787102 on Build GRCh38
rs545911521 Chr.10: 42787083-42787083 on Build GRCh38
rs748493682 Chr.10: 42787182-42787182 on Build GRCh38
rs918462638 Chr.10: 42787142-42787142 on Build GRCh38
rs761685443 Chr.10: 42787256-42787256 on Build GRCh38
rs1019595836 Chr.10: 42787101-42787101 on Build GRCh38
rs188002244 Chr.10: 42787081-42787081 on Build GRCh38
rs758651801 Chr.10: 42787172-42787172 on Build GRCh38

View Full Product Details