Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15485840-15486113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELA2B
Entrez Gene ID
51032
Gene Name
chymotrypsin like elastase family member 2B
Gene Aliases
ELA2B
UniGene
Hs.631871
Gene Chromosome Location
Chr.1: 15476101-15491400 on Build GRCh38
Transcripts
NM_015849.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6626077 c.573G>A p.W191* Chr.1: 15485980-15485980 on Build GRCh38
7163089 c.506T>C p.L169P Chr.1: 15485913-15485913 on Build GRCh38
7136401 c.594G>T p.T198T Chr.1: 15486001-15486001 on Build GRCh38
3802347 c.553A>C p.T185P Chr.1: 15485960-15485960 on Build GRCh38
5844082 c.505C>T p.L169F Chr.1: 15485912-15485912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764666444 Chr.1: 15486072-15486072 on Build GRCh38
rs768856027 Chr.1: 15486014-15486014 on Build GRCh38
rs898396980 Chr.1: 15485973-15485973 on Build GRCh38
rs780634627 Chr.1: 15486090-15486090 on Build GRCh38
rs534649662 Chr.1: 15486029-15486029 on Build GRCh38
rs757796070 Chr.1: 15485983-15485983 on Build GRCh38
rs768903430 Chr.1: 15485920-15485920 on Build GRCh38
rs780397964 Chr.1: 15485997-15485997 on Build GRCh38
rs770320071 Chr.1: 15485994-15485994 on Build GRCh38
rs746271233 Chr.1: 15485993-15485993 on Build GRCh38
rs1047298892 Chr.1: 15485977-15485977 on Build GRCh38
rs148560379 Chr.1: 15485905-15485905 on Build GRCh38
rs369557552 Chr.1: 15486081-15486081 on Build GRCh38
rs754469867 Chr.1: 15485959-15485959 on Build GRCh38
rs763092183 Chr.1: 15485909-15485909 on Build GRCh38
rs753531271 Chr.1: 15485955-15485955 on Build GRCh38
rs781622980 Chr.1: 15485900-15485900 on Build GRCh38
rs746018902 Chr.1: 15485904-15485904 on Build GRCh38
rs201761312 Chr.1: 15485946-15485946 on Build GRCh38
rs994025897 Chr.1: 15485974-15485974 on Build GRCh38
rs753482309 Chr.1: 15486046-15486046 on Build GRCh38
rs768179849 Chr.1: 15486030-15486031 on Build GRCh38
rs942639242 Chr.1: 15485943-15485955 on Build GRCh38
rs200907848 Chr.1: 15486015-15486015 on Build GRCh38
rs201229614 Chr.1: 15486071-15486071 on Build GRCh38
rs750246581 Chr.1: 15485944-15485944 on Build GRCh38
rs202010806 Chr.1: 15486047-15486047 on Build GRCh38
rs758864976 Chr.1: 15485879-15485879 on Build GRCh38
rs760490653 Chr.1: 15486031-15486031 on Build GRCh38
rs781701261 Chr.1: 15486083-15486083 on Build GRCh38
rs199863613 Chr.1: 15485906-15485906 on Build GRCh38
rs373761038 Chr.1: 15485869-15485869 on Build GRCh38
rs372520727 Chr.1: 15486086-15486086 on Build GRCh38
rs778460448 Chr.1: 15485981-15485981 on Build GRCh38
rs761880513 Chr.1: 15485933-15485933 on Build GRCh38
rs749582741 Chr.1: 15486000-15486000 on Build GRCh38
rs574028572 Chr.1: 15485995-15485995 on Build GRCh38
rs761598862 Chr.1: 15486021-15486021 on Build GRCh38
rs766212606 Chr.1: 15486043-15486043 on Build GRCh38
rs748820348 Chr.1: 15485871-15485871 on Build GRCh38
rs762475952 Chr.1: 15485937-15485937 on Build GRCh38
rs774420011 Chr.1: 15485925-15485925 on Build GRCh38
rs750939232 Chr.1: 15486087-15486087 on Build GRCh38
rs751943635 Chr.1: 15485982-15485982 on Build GRCh38
rs747571473 Chr.1: 15485884-15485884 on Build GRCh38
rs6429745 Chr.1: 15485937-15485937 on Build GRCh38
rs41270253 Chr.1: 15485868-15485868 on Build GRCh38
rs781358035 Chr.1: 15485992-15485992 on Build GRCh38
rs200058067 Chr.1: 15486028-15486028 on Build GRCh38
rs774581420 Chr.1: 15486018-15486018 on Build GRCh38
rs766052437 Chr.1: 15485954-15485954 on Build GRCh38
rs778223622 Chr.1: 15485883-15485883 on Build GRCh38
rs775210115 Chr.1: 15485913-15485913 on Build GRCh38
rs756532296 Chr.1: 15486089-15486089 on Build GRCh38

View Full Product Details