Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 111693672-111693903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf84
Entrez Gene ID
158401
Gene Name
chromosome 9 open reading frame 84
Gene Aliases
-
UniGene
Hs.428209
Gene Chromosome Location
Chr.9: 111686175-111795949 on Build GRCh38
Transcripts
NM_001080551.2 NM_173521.4 NR_109816.1 XM_006716974.2 XM_006716975.2 XM_011518302.2 XM_011518303.1 XM_011518306.1 XM_017014340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5752917 c.3227delG p.C1076fs*15 Chr.9: 111693845-111693845 on Build GRCh38
5752918 c.3110delG p.C1037fs*15 Chr.9: 111693845-111693845 on Build GRCh38
3735328 c.3182T>A p.L1061Q Chr.9: 111693890-111693890 on Build GRCh38
5522263 c.3220A>G p.T1074A Chr.9: 111693852-111693852 on Build GRCh38
5522264 c.3103A>G p.T1035A Chr.9: 111693852-111693852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762305471 Chr.9: 111693848-111693848 on Build GRCh38
rs199910267 Chr.9: 111693784-111693784 on Build GRCh38
rs747749499 Chr.9: 111693767-111693771 on Build GRCh38
rs1045999924 Chr.9: 111693729-111693729 on Build GRCh38
rs773570545 Chr.9: 111693864-111693864 on Build GRCh38
rs776994822 Chr.9: 111693840-111693840 on Build GRCh38
rs777553199 Chr.9: 111693761-111693761 on Build GRCh38
rs755853885 Chr.9: 111693759-111693759 on Build GRCh38
rs564781720 Chr.9: 111693822-111693822 on Build GRCh38
rs1028515519 Chr.9: 111693862-111693862 on Build GRCh38
rs562290041 Chr.9: 111693703-111693703 on Build GRCh38
rs186188808 Chr.9: 111693748-111693748 on Build GRCh38
rs765525411 Chr.9: 111693857-111693857 on Build GRCh38
rs142785470 Chr.9: 111693868-111693868 on Build GRCh38
rs893302291 Chr.9: 111693808-111693808 on Build GRCh38
rs903470639 Chr.9: 111693737-111693737 on Build GRCh38
rs572569210 Chr.9: 111693747-111693747 on Build GRCh38
rs746089963 Chr.9: 111693778-111693778 on Build GRCh38
rs1005843140 Chr.9: 111693847-111693847 on Build GRCh38
rs377312835 Chr.9: 111693752-111693752 on Build GRCh38
rs775747035 Chr.9: 111693795-111693795 on Build GRCh38
rs376244567 Chr.9: 111693811-111693811 on Build GRCh38
rs762537168 Chr.9: 111693865-111693865 on Build GRCh38

View Full Product Details