Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70240030-70240263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AWAT1
Entrez Gene ID
158833
Gene Name
acyl-CoA wax alcohol acyltransferase 1
Gene Aliases
DGA2, DGAT2L3
UniGene
Hs.407641
Gene Chromosome Location
Chr.X: 70233489-70240661 on Build GRCh38
Transcripts
NM_001013579.2 XM_017029295.1

SNP Detail

SNP ID Chromosome Location
rs953181845 Chr.X: 70240126-70240126 on Build GRCh38
rs977351579 Chr.X: 70240224-70240224 on Build GRCh38
rs914258566 Chr.X: 70240200-70240200 on Build GRCh38
rs778784686 Chr.X: 70240106-70240106 on Build GRCh38
rs1015708121 Chr.X: 70240093-70240093 on Build GRCh38
rs769336646 Chr.X: 70240225-70240225 on Build GRCh38
rs199722389 Chr.X: 70240182-70240182 on Build GRCh38
rs769985471 Chr.X: 70240173-70240173 on Build GRCh38
rs191188465 Chr.X: 70240121-70240121 on Build GRCh38
rs146722168 Chr.X: 70240233-70240233 on Build GRCh38
rs945717086 Chr.X: 70240203-70240203 on Build GRCh38
rs760331960 Chr.X: 70240234-70240234 on Build GRCh38
rs1152194 Chr.X: 70240095-70240095 on Build GRCh38
rs753667214 Chr.X: 70240089-70240089 on Build GRCh38
rs183658945 Chr.X: 70240136-70240136 on Build GRCh38
rs777803515 Chr.X: 70240128-70240128 on Build GRCh38
rs369158754 Chr.X: 70240122-70240122 on Build GRCh38
rs1013184952 Chr.X: 70240052-70240052 on Build GRCh38
rs186333842 Chr.X: 70240120-70240120 on Build GRCh38
rs752184357 Chr.X: 70240087-70240087 on Build GRCh38
rs1045978987 Chr.X: 70240072-70240072 on Build GRCh38
rs61750915 Chr.X: 70240176-70240176 on Build GRCh38
rs200229235 Chr.X: 70240149-70240149 on Build GRCh38
rs771136550 Chr.X: 70240137-70240137 on Build GRCh38
rs894765876 Chr.X: 70240048-70240048 on Build GRCh38
rs778111564 Chr.X: 70240138-70240138 on Build GRCh38
rs767145587 Chr.X: 70240084-70240084 on Build GRCh38
rs993730976 Chr.X: 70240076-70240076 on Build GRCh38

View Full Product Details