Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 121377464-121377725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLASP1
Entrez Gene ID
23332
Gene Name
cytoplasmic linker associated protein 1
Gene Aliases
MAST1
UniGene
Hs.469840
Gene Chromosome Location
Chr.2: 121337776-121649476 on Build GRCh38
Transcripts
NM_001142273.1 NM_001142274.1 NM_001207051.1 NM_015282.2 XM_006712377.1 XM_006712381.1 XM_006712382.1 XM_006712383.1 XM_006712385.1 XM_011510848.1 XM_011510849.1 XM_017003657.1 XM_017003658.1 XM_017003659.1 XM_017003660.1 XM_017003661.1 XM_017003662.1 XM_017003663.1 XM_017003664.1 XM_017003665.1 XM_017003666.1 XM_017003667.1 XM_017003668.1 XM_017003669.1 XM_017003670.1 XM_017003671.1 XM_017003672.1 XM_017003673.1 XM_017003674.1 XM_017003675.1 XM_017003676.1 XM_017003677.1 XM_017003678.1 XM_017003679.1 XM_017003680.1 XM_017003681.1 XM_017003682.1 XM_017003683.1 XM_017003684.1 XM_017003685.1 XM_017003686.1 XM_017003687.1 XM_017003688.1 XM_017003689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6509690 c.3407G>T p.S1136I Chr.2: 121377602-121377602 on Build GRCh38
6509689 c.3539G>T p.S1180I Chr.2: 121377602-121377602 on Build GRCh38
3701929 c.2649G>T p.L883L Chr.2: 121377643-121377643 on Build GRCh38
4809946 c.3483A>G p.K1161K Chr.2: 121377526-121377526 on Build GRCh38
5647861 c.2699G>A p.R900H Chr.2: 121377593-121377593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs578121908 Chr.2: 121377646-121377646 on Build GRCh38
rs775867411 Chr.2: 121377546-121377546 on Build GRCh38
rs372952192 Chr.2: 121377647-121377647 on Build GRCh38
rs769714674 Chr.2: 121377507-121377507 on Build GRCh38
rs767421642 Chr.2: 121377586-121377586 on Build GRCh38
rs370401387 Chr.2: 121377594-121377594 on Build GRCh38
rs747918004 Chr.2: 121377506-121377506 on Build GRCh38
rs772044172 Chr.2: 121377649-121377649 on Build GRCh38
rs762559034 Chr.2: 121377572-121377572 on Build GRCh38
rs775701014 Chr.2: 121377666-121377666 on Build GRCh38
rs759557730 Chr.2: 121377535-121377535 on Build GRCh38
rs903694985 Chr.2: 121377547-121377547 on Build GRCh38
rs777680280 Chr.2: 121377595-121377595 on Build GRCh38
rs749507352 Chr.2: 121377512-121377512 on Build GRCh38
rs772575850 Chr.2: 121377539-121377539 on Build GRCh38
rs888765139 Chr.2: 121377682-121377682 on Build GRCh38
rs751028676 Chr.2: 121377582-121377582 on Build GRCh38
rs774450164 Chr.2: 121377524-121377524 on Build GRCh38
rs548012509 Chr.2: 121377694-121377694 on Build GRCh38
rs777778172 Chr.2: 121377510-121377510 on Build GRCh38
rs757506553 Chr.2: 121377636-121377636 on Build GRCh38
rs749127142 Chr.2: 121377623-121377623 on Build GRCh38
rs754245466 Chr.2: 121377557-121377557 on Build GRCh38
rs948152640 Chr.2: 121377508-121377508 on Build GRCh38
rs752571395 Chr.2: 121377593-121377593 on Build GRCh38
rs116635626 Chr.2: 121377681-121377681 on Build GRCh38
rs766048276 Chr.2: 121377578-121377578 on Build GRCh38
rs375752731 Chr.2: 121377550-121377550 on Build GRCh38
rs771319840 Chr.2: 121377523-121377523 on Build GRCh38
rs754458736 Chr.2: 121377584-121377584 on Build GRCh38

View Full Product Details