Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 110210642-110210867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC40
Entrez Gene ID
51362
Gene Name
cell division cycle 40
Gene Aliases
EHB3, PRP17, PRPF17
UniGene
Hs.428147
Gene Chromosome Location
Chr.6: 110180421-110232220 on Build GRCh38
Transcripts
NM_015891.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3777007 c.652G>A p.E218K Chr.6: 110210728-110210728 on Build GRCh38
1072107 c.643G>T p.E215* Chr.6: 110210719-110210719 on Build GRCh38
7244171 c.727+4A>C p.? Chr.6: 110210807-110210807 on Build GRCh38
3777006 c.652G>A p.E218K Chr.6: 110210728-110210728 on Build GRCh38
1487114 c.666G>C p.K222N Chr.6: 110210742-110210742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759857434 Chr.6: 110210794-110210794 on Build GRCh38
rs929043494 Chr.6: 110210736-110210736 on Build GRCh38
rs751647289 Chr.6: 110210777-110210777 on Build GRCh38
rs746200903 Chr.6: 110210708-110210708 on Build GRCh38
rs776616990 Chr.6: 110210754-110210754 on Build GRCh38
rs775314336 Chr.6: 110210722-110210722 on Build GRCh38
rs201662538 Chr.6: 110210748-110210748 on Build GRCh38
rs745550749 Chr.6: 110210688-110210688 on Build GRCh38
rs756723109 Chr.6: 110210811-110210812 on Build GRCh38
rs577890969 Chr.6: 110210773-110210773 on Build GRCh38
rs760430736 Chr.6: 110210735-110210735 on Build GRCh38
rs746536048 Chr.6: 110210809-110210810 on Build GRCh38
rs761597070 Chr.6: 110210763-110210763 on Build GRCh38
rs771665751 Chr.6: 110210712-110210712 on Build GRCh38
rs777520857 Chr.6: 110210677-110210678 on Build GRCh38

View Full Product Details