Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202464708-202464933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMPR2
Entrez Gene ID
659
Gene Name
bone morphogenetic protein receptor type 2
Gene Aliases
BMPR-II, BMPR3, BMR2, BRK-3, POVD1, PPH1, T-ALK
UniGene
Hs.471119
Gene Chromosome Location
Chr.2: 202376310-202567751 on Build GRCh38
Transcripts
NM_001204.6 XM_011511687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6156563 c.181T>A p.S61T Chr.2: 202464913-202464913 on Build GRCh38
6612043 c.152_153delTC p.H53fs*1 Chr.2: 202464884-202464885 on Build GRCh38
4188939 c.193delA p.T65fs*13 Chr.2: 202464925-202464925 on Build GRCh38
6612047 c.105G>A p.A35A Chr.2: 202464837-202464837 on Build GRCh38
4090293 c.116C>T p.P39L Chr.2: 202464848-202464848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137852755 Chr.2: 202464852-202464852 on Build GRCh38
rs200976092 Chr.2: 202464897-202464897 on Build GRCh38
rs747740309 Chr.2: 202464821-202464821 on Build GRCh38
rs886039223 Chr.2: 202464907-202464908 on Build GRCh38
rs759804753 Chr.2: 202464868-202464868 on Build GRCh38
rs573463511 Chr.2: 202464812-202464812 on Build GRCh38
rs770804811 Chr.2: 202464848-202464848 on Build GRCh38
rs1051805297 Chr.2: 202464887-202464887 on Build GRCh38
rs764721647 Chr.2: 202464779-202464779 on Build GRCh38
rs754688935 Chr.2: 202464816-202464816 on Build GRCh38
rs752005716 Chr.2: 202464906-202464906 on Build GRCh38
rs368430622 Chr.2: 202464827-202464827 on Build GRCh38
rs147207234 Chr.2: 202464813-202464813 on Build GRCh38
rs770128128 Chr.2: 202464836-202464836 on Build GRCh38
rs757551343 Chr.2: 202464795-202464795 on Build GRCh38
rs912963620 Chr.2: 202464751-202464751 on Build GRCh38
rs112862820 Chr.2: 202464818-202464818 on Build GRCh38
rs375690035 Chr.2: 202464757-202464757 on Build GRCh38
rs201759998 Chr.2: 202464882-202464882 on Build GRCh38
rs751917686 Chr.2: 202464782-202464782 on Build GRCh38
rs776603012 Chr.2: 202464849-202464849 on Build GRCh38
rs752864146 Chr.2: 202464765-202464765 on Build GRCh38
rs746943205 Chr.2: 202464837-202464837 on Build GRCh38
rs376957186 Chr.2: 202464770-202464770 on Build GRCh38
rs945836014 Chr.2: 202464788-202464788 on Build GRCh38
rs771887212 Chr.2: 202464826-202464826 on Build GRCh38
rs150080314 Chr.2: 202464879-202464879 on Build GRCh38

View Full Product Details