Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100218222-100218479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR6
Entrez Gene ID
64431
Gene Name
ARP6 actin-related protein 6 homolog
Gene Aliases
ARP6, CDA12, HSPC281, MSTP136, hARP6, hARPX
UniGene
Hs.115088
Gene Chromosome Location
Chr.12: 100200087-100224425 on Build GRCh38
Transcripts
NM_022496.4 NR_048568.1 NR_048569.1 XM_017019821.1 NR_135073.1

Gene Symbol
DEPDC4
Entrez Gene ID
120863
Gene Name
DEP domain containing 4
Gene Aliases
DEP.4
UniGene
Hs.653118
Gene Chromosome Location
Chr.12: 100203669-100268094 on Build GRCh38
Transcripts
NM_022496.4 NR_048568.1 NR_048569.1 XM_017019821.1 NR_135073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3954022 c.790G>A p.G264R Chr.12: 100218454-100218454 on Build GRCh38
1660381 c.814A>G p.N272D Chr.12: 100218478-100218478 on Build GRCh38
4574924 c.807T>C p.R269R Chr.12: 100218471-100218471 on Build GRCh38
6438912 c.805C>T p.R269C Chr.12: 100218469-100218469 on Build GRCh38
6432941 c.779A>G p.K260R Chr.12: 100218443-100218443 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777006612 Chr.12: 100218400-100218400 on Build GRCh38
rs773401737 Chr.12: 100218427-100218427 on Build GRCh38
rs1014301162 Chr.12: 100218364-100218364 on Build GRCh38
rs895978939 Chr.12: 100218361-100218361 on Build GRCh38
rs1055958070 Chr.12: 100218318-100218318 on Build GRCh38
rs946401455 Chr.12: 100218280-100218280 on Build GRCh38
rs1002233671 Chr.12: 100218303-100218303 on Build GRCh38
rs780680626 Chr.12: 100218369-100218369 on Build GRCh38
rs747478178 Chr.12: 100218375-100218375 on Build GRCh38
rs757842084 Chr.12: 100218365-100218366 on Build GRCh38
rs762125823 Chr.12: 100218408-100218408 on Build GRCh38
rs1043402650 Chr.12: 100218286-100218286 on Build GRCh38
rs770044278 Chr.12: 100218416-100218416 on Build GRCh38
rs371505912 Chr.12: 100218431-100218431 on Build GRCh38
rs531524022 Chr.12: 100218314-100218314 on Build GRCh38
rs1052618628 Chr.12: 100218268-100218268 on Build GRCh38
rs766364029 Chr.12: 100218432-100218432 on Build GRCh38
rs764421808 Chr.12: 100218270-100218270 on Build GRCh38
rs184875952 Chr.12: 100218287-100218287 on Build GRCh38
rs769151827 Chr.12: 100218380-100218380 on Build GRCh38

View Full Product Details