Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73740201-73740468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf59
Entrez Gene ID
388135
Gene Name
chromosome 15 open reading frame 59
Gene Aliases
-
UniGene
Hs.213493
Gene Chromosome Location
Chr.15: 73735431-73753170 on Build GRCh38
Transcripts
NM_001039614.2 NM_001303254.1 XM_005254369.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7358091 c.509C>T p.T170M Chr.15: 73740290-73740290 on Build GRCh38
127468 c.510G>T p.T170T Chr.15: 73740289-73740289 on Build GRCh38
1165486 c.533G>A p.R178Q Chr.15: 73740266-73740266 on Build GRCh38
6001673 c.448A>G p.N150D Chr.15: 73740351-73740351 on Build GRCh38
4056751 c.401G>A p.G134E Chr.15: 73740398-73740398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773537559 Chr.15: 73740240-73740240 on Build GRCh38
rs144142309 Chr.15: 73740437-73740437 on Build GRCh38
rs549585440 Chr.15: 73740302-73740302 on Build GRCh38
rs751144619 Chr.15: 73740358-73740358 on Build GRCh38
rs372039755 Chr.15: 73740362-73740362 on Build GRCh38
rs755975082 Chr.15: 73740381-73740381 on Build GRCh38
rs901896697 Chr.15: 73740340-73740340 on Build GRCh38
rs577646174 Chr.15: 73740360-73740360 on Build GRCh38
rs942515888 Chr.15: 73740328-73740328 on Build GRCh38
rs753836298 Chr.15: 73740426-73740426 on Build GRCh38
rs780211009 Chr.15: 73740388-73740388 on Build GRCh38
rs756101671 Chr.15: 73740289-73740289 on Build GRCh38
rs762561178 Chr.15: 73740263-73740263 on Build GRCh38
rs747470845 Chr.15: 73740415-73740415 on Build GRCh38
rs748488374 Chr.15: 73740258-73740258 on Build GRCh38
rs774974645 Chr.15: 73740341-73740341 on Build GRCh38
rs750283431 Chr.15: 73740380-73740380 on Build GRCh38
rs755055161 Chr.15: 73740404-73740404 on Build GRCh38
rs1040912400 Chr.15: 73740441-73740441 on Build GRCh38
rs764600744 Chr.15: 73740251-73740251 on Build GRCh38
rs754985503 Chr.15: 73740448-73740448 on Build GRCh38
rs906921128 Chr.15: 73740229-73740229 on Build GRCh38
rs752251135 Chr.15: 73740253-73740253 on Build GRCh38
rs768058085 Chr.15: 73740238-73740238 on Build GRCh38
rs767168787 Chr.15: 73740375-73740375 on Build GRCh38
rs773873738 Chr.15: 73740433-73740433 on Build GRCh38
rs773644368 Chr.15: 73740322-73740322 on Build GRCh38
rs764736416 Chr.15: 73740376-73740376 on Build GRCh38
rs970631920 Chr.15: 73740407-73740407 on Build GRCh38
rs777064188 Chr.15: 73740416-73740416 on Build GRCh38
rs1026505777 Chr.15: 73740280-73740280 on Build GRCh38
rs753956163 Chr.15: 73740389-73740389 on Build GRCh38
rs959807787 Chr.15: 73740413-73740413 on Build GRCh38
rs988449500 Chr.15: 73740241-73740241 on Build GRCh38
rs375165319 Chr.15: 73740434-73740434 on Build GRCh38
rs200967826 Chr.15: 73740410-73740410 on Build GRCh38
rs149521207 Chr.15: 73740427-73740427 on Build GRCh38
rs772281429 Chr.15: 73740320-73740320 on Build GRCh38
rs749423301 Chr.15: 73740294-73740294 on Build GRCh38
rs942540767 Chr.15: 73740438-73740438 on Build GRCh38
rs565532541 Chr.15: 73740414-73740414 on Build GRCh38
rs534540529 Chr.15: 73740319-73740319 on Build GRCh38
rs762339433 Chr.15: 73740221-73740221 on Build GRCh38
rs372773637 Chr.15: 73740411-73740411 on Build GRCh38
rs781046669 Chr.15: 73740283-73740283 on Build GRCh38
rs761221274 Chr.15: 73740350-73740350 on Build GRCh38
rs993231197 Chr.15: 73740260-73740260 on Build GRCh38
rs79475978 Chr.15: 73740290-73740290 on Build GRCh38
rs761273404 Chr.15: 73740243-73740243 on Build GRCh38
rs28594180 Chr.15: 73740442-73740442 on Build GRCh38
rs377764222 Chr.15: 73740267-73740267 on Build GRCh38
rs368003272 Chr.15: 73740308-73740308 on Build GRCh38
rs141142512 Chr.15: 73740447-73740447 on Build GRCh38
rs371495958 Chr.15: 73740331-73740331 on Build GRCh38
rs202072343 Chr.15: 73740266-73740266 on Build GRCh38
rs138123176 Chr.15: 73740405-73740405 on Build GRCh38
rs760578731 Chr.15: 73740439-73740439 on Build GRCh38
rs750288109 Chr.15: 73740286-73740286 on Build GRCh38
rs201933736 Chr.15: 73740384-73740384 on Build GRCh38
rs756987412 Chr.15: 73740271-73740271 on Build GRCh38
rs771505477 Chr.15: 73740334-73740334 on Build GRCh38

View Full Product Details