Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152719866-152720133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf68
Entrez Gene ID
100129271
Gene Name
chromosome 1 open reading frame 68
Gene Aliases
LEP7, XP32
UniGene
Hs.601077
Gene Chromosome Location
Chr.1: 152719522-152720429 on Build GRCh38
Transcripts
NM_001024679.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3771456 c.610T>C p.S204P Chr.1: 152720131-152720131 on Build GRCh38
3750362 c.515G>A p.C172Y Chr.1: 152720036-152720036 on Build GRCh38
5886024 c.587G>A p.G196E Chr.1: 152720108-152720108 on Build GRCh38
4409087 c.436C>T p.Q146* Chr.1: 152719957-152719957 on Build GRCh38
6215702 c.535G>A p.G179R Chr.1: 152720056-152720056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752481746 Chr.1: 152719913-152719913 on Build GRCh38
rs868304339 Chr.1: 152720026-152720026 on Build GRCh38
rs368436282 Chr.1: 152720039-152720039 on Build GRCh38
rs567419709 Chr.1: 152720075-152720075 on Build GRCh38
rs780408651 Chr.1: 152719960-152719960 on Build GRCh38
rs373807207 Chr.1: 152719958-152719958 on Build GRCh38
rs755843513 Chr.1: 152719910-152719910 on Build GRCh38
rs571448810 Chr.1: 152720048-152720048 on Build GRCh38
rs906549758 Chr.1: 152719946-152719946 on Build GRCh38
rs773269913 Chr.1: 152720005-152720009 on Build GRCh38
rs753395174 Chr.1: 152719914-152719914 on Build GRCh38
rs59194678 Chr.1: 152720038-152720038 on Build GRCh38
rs981936553 Chr.1: 152720030-152720030 on Build GRCh38
rs963558878 Chr.1: 152719990-152719990 on Build GRCh38
rs553777962 Chr.1: 152720074-152720074 on Build GRCh38
rs186816860 Chr.1: 152720012-152720012 on Build GRCh38
rs868766653 Chr.1: 152719921-152719921 on Build GRCh38
rs533918314 Chr.1: 152720058-152720058 on Build GRCh38
rs770154522 Chr.1: 152720043-152720043 on Build GRCh38
rs376518806 Chr.1: 152719941-152719941 on Build GRCh38
rs372420974 Chr.1: 152720087-152720087 on Build GRCh38
rs771012801 Chr.1: 152720056-152720056 on Build GRCh38
rs971477212 Chr.1: 152720025-152720025 on Build GRCh38
rs1009566350 Chr.1: 152719923-152719923 on Build GRCh38
rs41268474 Chr.1: 152720036-152720036 on Build GRCh38
rs1026564860 Chr.1: 152720007-152720007 on Build GRCh38
rs1033256715 Chr.1: 152719993-152719993 on Build GRCh38
rs987730232 Chr.1: 152720021-152720021 on Build GRCh38
rs370737663 Chr.1: 152719947-152719947 on Build GRCh38
rs944682 Chr.1: 152720112-152720112 on Build GRCh38
rs760632549 Chr.1: 152720055-152720055 on Build GRCh38
rs191594967 Chr.1: 152720047-152720047 on Build GRCh38
rs1002215238 Chr.1: 152719970-152719970 on Build GRCh38
rs908373091 Chr.1: 152720031-152720031 on Build GRCh38
rs873775 Chr.1: 152719996-152719996 on Build GRCh38
rs1033743452 Chr.1: 152719986-152719986 on Build GRCh38
rs756929875 Chr.1: 152719917-152719917 on Build GRCh38

View Full Product Details