Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26445657-26445916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
XM_011514268.2 NM_001242803.1 NM_006994.4 NM_197974.2

Gene Symbol
BTN3A3
Entrez Gene ID
10384
Gene Name
butyrophilin subfamily 3 member A3
Gene Aliases
BTF3, BTN3.3
UniGene
Hs.167741
Gene Chromosome Location
Chr.6: 26440472-26453415 on Build GRCh38
Transcripts
XM_011514268.2 NM_001242803.1 NM_006994.4 NM_197974.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1076800 c.443C>A p.S148Y Chr.6: 26445713-26445713 on Build GRCh38
6497922 c.498C>T p.C166C Chr.6: 26445768-26445768 on Build GRCh38
7421466 c.515A>T p.Y172F Chr.6: 26445785-26445785 on Build GRCh38
1076801 c.540C>T p.S180S Chr.6: 26445810-26445810 on Build GRCh38
318963 c.549G>T p.K183N Chr.6: 26445819-26445819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912956996 Chr.6: 26445700-26445700 on Build GRCh38
rs780652944 Chr.6: 26445714-26445714 on Build GRCh38
rs751053710 Chr.6: 26445703-26445703 on Build GRCh38
rs761543235 Chr.6: 26445895-26445895 on Build GRCh38
rs749042680 Chr.6: 26445742-26445742 on Build GRCh38
rs759929819 Chr.6: 26445877-26445877 on Build GRCh38
rs749936162 Chr.6: 26445744-26445751 on Build GRCh38
rs138065289 Chr.6: 26445783-26445783 on Build GRCh38
rs150636780 Chr.6: 26445786-26445786 on Build GRCh38
rs373589611 Chr.6: 26445793-26445793 on Build GRCh38
rs570459961 Chr.6: 26445858-26445858 on Build GRCh38
rs754962015 Chr.6: 26445725-26445725 on Build GRCh38
rs773420672 Chr.6: 26445784-26445784 on Build GRCh38
rs748293471 Chr.6: 26445780-26445780 on Build GRCh38
rs777169027 Chr.6: 26445791-26445791 on Build GRCh38
rs911431072 Chr.6: 26445749-26445749 on Build GRCh38
rs778010971 Chr.6: 26445869-26445869 on Build GRCh38
rs770264493 Chr.6: 26445884-26445884 on Build GRCh38
rs774580341 Chr.6: 26445756-26445756 on Build GRCh38
rs755788691 Chr.6: 26445750-26445751 on Build GRCh38
rs148026276 Chr.6: 26445728-26445728 on Build GRCh38
rs745877467 Chr.6: 26445715-26445715 on Build GRCh38
rs754846329 Chr.6: 26445846-26445846 on Build GRCh38
rs541415098 Chr.6: 26445870-26445870 on Build GRCh38
rs929963506 Chr.6: 26445797-26445797 on Build GRCh38
rs749879449 Chr.6: 26445833-26445833 on Build GRCh38
rs779840810 Chr.6: 26445734-26445734 on Build GRCh38
rs748468450 Chr.6: 26445857-26445857 on Build GRCh38
rs767075667 Chr.6: 26445832-26445832 on Build GRCh38
rs766138602 Chr.6: 26445819-26445819 on Build GRCh38
rs986912157 Chr.6: 26445740-26445740 on Build GRCh38
rs766933059 Chr.6: 26445788-26445788 on Build GRCh38
rs201388694 Chr.6: 26445838-26445838 on Build GRCh38
rs771589705 Chr.6: 26445875-26445875 on Build GRCh38
rs756840647 Chr.6: 26445705-26445705 on Build GRCh38
rs920188754 Chr.6: 26445794-26445794 on Build GRCh38
rs765497042 Chr.6: 26445810-26445810 on Build GRCh38
rs377686214 Chr.6: 26445834-26445834 on Build GRCh38
rs768788537 Chr.6: 26445746-26445746 on Build GRCh38
rs965704959 Chr.6: 26445820-26445820 on Build GRCh38
rs778536905 Chr.6: 26445847-26445847 on Build GRCh38
rs777245190 Chr.6: 26445876-26445876 on Build GRCh38
rs767093669 Chr.6: 26445896-26445896 on Build GRCh38
rs951920502 Chr.6: 26445841-26445841 on Build GRCh38
rs763764002 Chr.6: 26445696-26445696 on Build GRCh38
rs374130285 Chr.6: 26445829-26445829 on Build GRCh38
rs751265742 Chr.6: 26445822-26445822 on Build GRCh38
rs775709582 Chr.6: 26445889-26445889 on Build GRCh38

View Full Product Details