Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68866003-68866238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP10
Entrez Gene ID
27302
Gene Name
bone morphogenetic protein 10
Gene Aliases
-
UniGene
Hs.158317
Gene Chromosome Location
Chr.2: 68865481-68871517 on Build GRCh38
Transcripts
NM_014482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3055874 c.738G>T p.Q246H Chr.2: 68866168-68866168 on Build GRCh38
4978491 c.819G>A p.M273I Chr.2: 68866087-68866087 on Build GRCh38
394501 c.801G>A p.K267K Chr.2: 68866105-68866105 on Build GRCh38
3055871 c.765G>A p.V255V Chr.2: 68866141-68866141 on Build GRCh38
3939136 c.712C>T p.R238W Chr.2: 68866194-68866194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144469983 Chr.2: 68866194-68866194 on Build GRCh38
rs998475946 Chr.2: 68866040-68866040 on Build GRCh38
rs555314842 Chr.2: 68866050-68866050 on Build GRCh38
rs2231344 Chr.2: 68866180-68866180 on Build GRCh38
rs746919370 Chr.2: 68866202-68866202 on Build GRCh38
rs949047379 Chr.2: 68866140-68866140 on Build GRCh38
rs1030346192 Chr.2: 68866042-68866042 on Build GRCh38
rs903410373 Chr.2: 68866157-68866157 on Build GRCh38
rs2231346 Chr.2: 68866123-68866123 on Build GRCh38
rs758292306 Chr.2: 68866199-68866199 on Build GRCh38
rs765578475 Chr.2: 68866102-68866102 on Build GRCh38
rs764620390 Chr.2: 68866144-68866144 on Build GRCh38
rs112193833 Chr.2: 68866081-68866081 on Build GRCh38
rs760130579 Chr.2: 68866059-68866059 on Build GRCh38
rs777738014 Chr.2: 68866201-68866201 on Build GRCh38
rs754104216 Chr.2: 68866172-68866172 on Build GRCh38
rs2231345 Chr.2: 68866156-68866156 on Build GRCh38
rs772741994 Chr.2: 68866039-68866039 on Build GRCh38
rs753046138 Chr.2: 68866120-68866120 on Build GRCh38
rs771981507 Chr.2: 68866026-68866026 on Build GRCh38
rs957479756 Chr.2: 68866191-68866191 on Build GRCh38
rs759221950 Chr.2: 68866098-68866098 on Build GRCh38
rs778579679 Chr.2: 68866193-68866193 on Build GRCh38
rs776306538 Chr.2: 68866083-68866083 on Build GRCh38
rs572462822 Chr.2: 68866126-68866126 on Build GRCh38
rs564324708 Chr.2: 68866176-68866176 on Build GRCh38
rs371768311 Chr.2: 68866213-68866213 on Build GRCh38
rs751708681 Chr.2: 68866149-68866149 on Build GRCh38
rs770494705 Chr.2: 68866209-68866209 on Build GRCh38
rs776105366 Chr.2: 68866214-68866214 on Build GRCh38
rs770406675 Chr.2: 68866072-68866072 on Build GRCh38
rs867162659 Chr.2: 68866101-68866101 on Build GRCh38

View Full Product Details