Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 224774962-224775203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK10
Entrez Gene ID
55619
Gene Name
dedicator of cytokinesis 10
Gene Aliases
DRIP2, Nbla10300, ZIZ3
UniGene
Hs.46578
Gene Chromosome Location
Chr.2: 224765090-225042689 on Build GRCh38
Transcripts
NM_001290263.1 NM_014689.2 XM_005246673.4 XM_006712617.3 XM_017004441.1 XM_017004442.1 XM_017004443.1 XM_017004444.1 XM_017004445.1 XM_017004446.1 XM_017004447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1405871 c.1458C>T p.F486F Chr.2: 224774981-224774981 on Build GRCh38
4501562 c.1447C>T p.R483C Chr.2: 224774992-224774992 on Build GRCh38
7324344 c.1400A>G p.E467G Chr.2: 224775039-224775039 on Build GRCh38
6157290 c.5803C>A p.P1935T Chr.2: 224775109-224775109 on Build GRCh38
4553428 c.1426G>A p.E476K Chr.2: 224775013-224775013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34478767 Chr.2: 224775067-224775068 on Build GRCh38
rs746885138 Chr.2: 224775065-224775065 on Build GRCh38
rs375646049 Chr.2: 224774980-224774980 on Build GRCh38
rs147390351 Chr.2: 224775160-224775160 on Build GRCh38
rs754737135 Chr.2: 224775040-224775040 on Build GRCh38
rs750844764 Chr.2: 224775016-224775016 on Build GRCh38
rs774532728 Chr.2: 224775100-224775100 on Build GRCh38
rs186781495 Chr.2: 224775172-224775172 on Build GRCh38
rs1049976530 Chr.2: 224775009-224775009 on Build GRCh38
rs771744068 Chr.2: 224775057-224775057 on Build GRCh38
rs764284194 Chr.2: 224774991-224774991 on Build GRCh38
rs778705314 Chr.2: 224775052-224775052 on Build GRCh38
rs895214108 Chr.2: 224775104-224775104 on Build GRCh38
rs777792020 Chr.2: 224775060-224775060 on Build GRCh38
rs924679265 Chr.2: 224775178-224775178 on Build GRCh38
rs367785024 Chr.2: 224775056-224775056 on Build GRCh38
rs368630062 Chr.2: 224775111-224775111 on Build GRCh38
rs775600520 Chr.2: 224775070-224775070 on Build GRCh38
rs888288804 Chr.2: 224775059-224775059 on Build GRCh38
rs755681454 Chr.2: 224775027-224775027 on Build GRCh38
rs538442939 Chr.2: 224775164-224775164 on Build GRCh38
rs767733062 Chr.2: 224775147-224775147 on Build GRCh38
rs768903758 Chr.2: 224775093-224775093 on Build GRCh38
rs374596718 Chr.2: 224775110-224775110 on Build GRCh38
rs1008538331 Chr.2: 224775034-224775034 on Build GRCh38
rs762258455 Chr.2: 224775101-224775101 on Build GRCh38
rs763012558 Chr.2: 224775083-224775083 on Build GRCh38
rs779364831 Chr.2: 224775028-224775028 on Build GRCh38
rs185653208 Chr.2: 224774981-224774981 on Build GRCh38
rs375579674 Chr.2: 224775153-224775153 on Build GRCh38
rs768117264 Chr.2: 224775014-224775014 on Build GRCh38
rs551976417 Chr.2: 224775171-224775171 on Build GRCh38
rs757608796 Chr.2: 224775005-224775005 on Build GRCh38
rs1038231747 Chr.2: 224775077-224775077 on Build GRCh38
rs16866167 Chr.2: 224775068-224775068 on Build GRCh38
rs374538454 Chr.2: 224775037-224775037 on Build GRCh38
rs757584142 Chr.2: 224774999-224774999 on Build GRCh38
rs372056146 Chr.2: 224775116-224775116 on Build GRCh38
rs761030588 Chr.2: 224775161-224775161 on Build GRCh38
rs370524123 Chr.2: 224774992-224774992 on Build GRCh38

View Full Product Details