Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36493584-36493845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXRED2
Entrez Gene ID
80020
Gene Name
FAD dependent oxidoreductase domain containing 2
Gene Aliases
ERFAD
UniGene
Hs.387601
Gene Chromosome Location
Chr.22: 36487186-36507101 on Build GRCh38
Transcripts
NM_001102371.1 NM_024955.5 XM_017028941.1 XM_017028942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5647379 c.1781G>A p.R594Q Chr.22: 36493647-36493647 on Build GRCh38
5631306 c.1780C>T p.R594* Chr.22: 36493648-36493648 on Build GRCh38
419567 c.1720C>T p.P574S Chr.22: 36493708-36493708 on Build GRCh38
419568 c.1721C>T p.P574L Chr.22: 36493707-36493707 on Build GRCh38
1682244 c.1693G>T p.E565* Chr.22: 36493735-36493735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202043879 Chr.22: 36493739-36493739 on Build GRCh38
rs778126816 Chr.22: 36493681-36493681 on Build GRCh38
rs771103235 Chr.22: 36493693-36493693 on Build GRCh38
rs765247209 Chr.22: 36493631-36493631 on Build GRCh38
rs370987188 Chr.22: 36493777-36493777 on Build GRCh38
rs762725592 Chr.22: 36493722-36493722 on Build GRCh38
rs771192890 Chr.22: 36493606-36493606 on Build GRCh38
rs957702707 Chr.22: 36493658-36493658 on Build GRCh38
rs776530236 Chr.22: 36493697-36493697 on Build GRCh38
rs780707318 Chr.22: 36493674-36493677 on Build GRCh38
rs942021784 Chr.22: 36493776-36493776 on Build GRCh38
rs928849614 Chr.22: 36493720-36493720 on Build GRCh38
rs200348211 Chr.22: 36493754-36493754 on Build GRCh38
rs755657123 Chr.22: 36493756-36493756 on Build GRCh38
rs763926357 Chr.22: 36493738-36493738 on Build GRCh38
rs776688131 Chr.22: 36493608-36493608 on Build GRCh38
rs921268070 Chr.22: 36493657-36493657 on Build GRCh38
rs756741928 Chr.22: 36493647-36493647 on Build GRCh38
rs751367817 Chr.22: 36493611-36493611 on Build GRCh38
rs758810388 Chr.22: 36493768-36493768 on Build GRCh38
rs147772253 Chr.22: 36493706-36493706 on Build GRCh38
rs371503116 Chr.22: 36493702-36493702 on Build GRCh38
rs756751585 Chr.22: 36493604-36493604 on Build GRCh38
rs747282570 Chr.22: 36493691-36493691 on Build GRCh38
rs1013689523 Chr.22: 36493793-36493793 on Build GRCh38
rs748319420 Chr.22: 36493761-36493761 on Build GRCh38
rs201122263 Chr.22: 36493785-36493785 on Build GRCh38
rs762816999 Chr.22: 36493638-36493638 on Build GRCh38
rs769976435 Chr.22: 36493812-36493812 on Build GRCh38
rs557756197 Chr.22: 36493755-36493755 on Build GRCh38
rs183299325 Chr.22: 36493618-36493618 on Build GRCh38
rs759427011 Chr.22: 36493615-36493615 on Build GRCh38
rs147607086 Chr.22: 36493662-36493662 on Build GRCh38
rs775500624 Chr.22: 36493707-36493707 on Build GRCh38
rs753301653 Chr.22: 36493762-36493762 on Build GRCh38
rs143414578 Chr.22: 36493774-36493774 on Build GRCh38
rs770361400 Chr.22: 36493721-36493721 on Build GRCh38
rs780706407 Chr.22: 36493773-36493774 on Build GRCh38
rs747409045 Chr.22: 36493775-36493775 on Build GRCh38
rs368642443 Chr.22: 36493795-36493795 on Build GRCh38
rs144263148 Chr.22: 36493700-36493700 on Build GRCh38
rs761419661 Chr.22: 36493741-36493741 on Build GRCh38
rs746393440 Chr.22: 36493707-36493707 on Build GRCh38
rs775095903 Chr.22: 36493633-36493633 on Build GRCh38
rs555593716 Chr.22: 36493680-36493680 on Build GRCh38
rs573196264 Chr.22: 36493786-36493786 on Build GRCh38
rs779834311 Chr.22: 36493814-36493814 on Build GRCh38
rs566658757 Chr.22: 36493645-36493645 on Build GRCh38
rs138229716 Chr.22: 36493663-36493663 on Build GRCh38
rs749714886 Chr.22: 36493814-36493815 on Build GRCh38
rs769559279 Chr.22: 36493703-36493703 on Build GRCh38
rs939828791 Chr.22: 36493746-36493746 on Build GRCh38
rs748396057 Chr.22: 36493666-36493666 on Build GRCh38
rs138563338 Chr.22: 36493769-36493769 on Build GRCh38
rs749980868 Chr.22: 36493661-36493661 on Build GRCh38
rs1031729766 Chr.22: 36493627-36493627 on Build GRCh38
rs542057393 Chr.22: 36493822-36493822 on Build GRCh38
rs566752752 Chr.22: 36493640-36493640 on Build GRCh38
rs755555284 Chr.22: 36493654-36493654 on Build GRCh38

View Full Product Details