Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76617494-76617725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2RL2
Entrez Gene ID
2151
Gene Name
coagulation factor II thrombin receptor like 2
Gene Aliases
PAR-3, PAR3
UniGene
Hs.42502
Gene Chromosome Location
Chr.5: 76615482-76623434 on Build GRCh38
Transcripts
NM_001256566.1 NM_004101.3 NM_001285460.1 NM_001285461.1 NM_001285462.1 NM_006633.3 XM_005248410.2 XM_005248414.1 XM_011543108.1 XM_017008960.1

Gene Symbol
IQGAP2
Entrez Gene ID
10788
Gene Name
IQ motif containing GTPase activating protein 2
Gene Aliases
-
UniGene
Hs.291030
Gene Chromosome Location
Chr.5: 76403255-76708132 on Build GRCh38
Transcripts
NM_001256566.1 NM_004101.3 NM_001285460.1 NM_001285461.1 NM_001285462.1 NM_006633.3 XM_005248410.2 XM_005248414.1 XM_011543108.1 XM_017008960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1620555 c.1074T>C p.Y358Y Chr.5: 76617633-76617633 on Build GRCh38
1567602 c.1076T>G p.F359C Chr.5: 76617631-76617631 on Build GRCh38
6900973 c.1043G>A p.S348N Chr.5: 76617664-76617664 on Build GRCh38
3073908 c.1050T>C p.N350N Chr.5: 76617657-76617657 on Build GRCh38
4155898 c.988T>C p.Y330H Chr.5: 76617719-76617719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867510403 Chr.5: 76617613-76617613 on Build GRCh38
rs779249772 Chr.5: 76617655-76617655 on Build GRCh38
rs2069684 Chr.5: 76617576-76617576 on Build GRCh38
rs772227689 Chr.5: 76617690-76617690 on Build GRCh38
rs990353044 Chr.5: 76617679-76617679 on Build GRCh38
rs777973383 Chr.5: 76617691-76617691 on Build GRCh38
rs749888207 Chr.5: 76617589-76617589 on Build GRCh38
rs542832114 Chr.5: 76617584-76617584 on Build GRCh38
rs769307910 Chr.5: 76617537-76617538 on Build GRCh38
rs866069607 Chr.5: 76617687-76617687 on Build GRCh38
rs748572943 Chr.5: 76617667-76617667 on Build GRCh38
rs762756940 Chr.5: 76617559-76617559 on Build GRCh38
rs572690258 Chr.5: 76617525-76617525 on Build GRCh38
rs775317252 Chr.5: 76617550-76617550 on Build GRCh38
rs763773916 Chr.5: 76617563-76617563 on Build GRCh38
rs755553172 Chr.5: 76617649-76617649 on Build GRCh38
rs777253204 Chr.5: 76617547-76617548 on Build GRCh38
rs767335456 Chr.5: 76617585-76617585 on Build GRCh38
rs2069701 Chr.5: 76617541-76617541 on Build GRCh38
rs755613069 Chr.5: 76617604-76617604 on Build GRCh38
rs367911000 Chr.5: 76617616-76617616 on Build GRCh38
rs199669532 Chr.5: 76617645-76617645 on Build GRCh38
rs958682642 Chr.5: 76617518-76617518 on Build GRCh38
rs200098123 Chr.5: 76617540-76617540 on Build GRCh38
rs762124428 Chr.5: 76617593-76617593 on Build GRCh38

View Full Product Details