Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67161741-67161994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL8
Entrez Gene ID
55336
Gene Name
F-box and leucine rich repeat protein 8
Gene Aliases
FBL8
UniGene
Hs.710714
Gene Chromosome Location
Chr.16: 67159988-67164174 on Build GRCh38
Transcripts
NM_018378.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5797383 c.98G>A p.R33K Chr.16: 67161883-67161883 on Build GRCh38
5740680 c.107C>T p.A36V Chr.16: 67161892-67161892 on Build GRCh38
1302111 c.48C>T p.I16I Chr.16: 67161833-67161833 on Build GRCh38
5595799 c.83C>T p.A28V Chr.16: 67161868-67161868 on Build GRCh38
5971854 c.6C>G p.A2A Chr.16: 67161791-67161791 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1004361548 Chr.16: 67161959-67161959 on Build GRCh38
rs141599444 Chr.16: 67161805-67161805 on Build GRCh38
rs544683763 Chr.16: 67161852-67161852 on Build GRCh38
rs375025400 Chr.16: 67161824-67161824 on Build GRCh38
rs117575136 Chr.16: 67161939-67161939 on Build GRCh38
rs765134800 Chr.16: 67161786-67161786 on Build GRCh38
rs142827853 Chr.16: 67161869-67161869 on Build GRCh38
rs374654506 Chr.16: 67161901-67161901 on Build GRCh38
rs779412523 Chr.16: 67161938-67161938 on Build GRCh38
rs932651101 Chr.16: 67161943-67161943 on Build GRCh38
rs564496926 Chr.16: 67161930-67161930 on Build GRCh38
rs936502487 Chr.16: 67161771-67161771 on Build GRCh38
rs1027059333 Chr.16: 67161917-67161917 on Build GRCh38
rs1002389252 Chr.16: 67161889-67161889 on Build GRCh38
rs747018340 Chr.16: 67161970-67161970 on Build GRCh38
rs138440769 Chr.16: 67161860-67161860 on Build GRCh38
rs112559086 Chr.16: 67161839-67161839 on Build GRCh38
rs371999405 Chr.16: 67161964-67161964 on Build GRCh38
rs774616423 Chr.16: 67161766-67161766 on Build GRCh38
rs763575991 Chr.16: 67161761-67161761 on Build GRCh38
rs773292555 Chr.16: 67161927-67161927 on Build GRCh38
rs761948952 Chr.16: 67161767-67161767 on Build GRCh38
rs1027775162 Chr.16: 67161863-67161863 on Build GRCh38
rs146097920 Chr.16: 67161884-67161884 on Build GRCh38
rs772199181 Chr.16: 67161905-67161905 on Build GRCh38
rs892358418 Chr.16: 67161828-67161828 on Build GRCh38
rs372208643 Chr.16: 67161915-67161915 on Build GRCh38
rs745589248 Chr.16: 67161825-67161825 on Build GRCh38
rs767861378 Chr.16: 67161770-67161770 on Build GRCh38
rs780519334 Chr.16: 67161838-67161838 on Build GRCh38
rs749881699 Chr.16: 67161902-67161902 on Build GRCh38
rs762606235 Chr.16: 67161880-67161880 on Build GRCh38
rs755226451 Chr.16: 67161924-67161924 on Build GRCh38
rs951056129 Chr.16: 67161854-67161854 on Build GRCh38
rs757183438 Chr.16: 67161896-67161896 on Build GRCh38
rs531038156 Chr.16: 67161803-67161803 on Build GRCh38
rs1046417194 Chr.16: 67161848-67161848 on Build GRCh38
rs751432584 Chr.16: 67161894-67161894 on Build GRCh38
rs371103065 Chr.16: 67161783-67161783 on Build GRCh38
rs35959876 Chr.16: 67161762-67161763 on Build GRCh38
rs774953768 Chr.16: 67161841-67161841 on Build GRCh38
rs562262960 Chr.16: 67161794-67161794 on Build GRCh38
rs755833140 Chr.16: 67161911-67161911 on Build GRCh38
rs113936384 Chr.16: 67161795-67161795 on Build GRCh38
rs750603635 Chr.16: 67161773-67161773 on Build GRCh38
rs751466702 Chr.16: 67161797-67161797 on Build GRCh38
rs769319847 Chr.16: 67161837-67161837 on Build GRCh38
rs968329339 Chr.16: 67161919-67161919 on Build GRCh38
rs150449492 Chr.16: 67161843-67161843 on Build GRCh38
rs769025234 Chr.16: 67161762-67161762 on Build GRCh38
rs761003885 Chr.16: 67161866-67161866 on Build GRCh38
rs756699436 Chr.16: 67161811-67161811 on Build GRCh38
rs780828331 Chr.16: 67161812-67161812 on Build GRCh38
rs768753697 Chr.16: 67161870-67161871 on Build GRCh38

View Full Product Details