Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101916040-101916295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUX1
Entrez Gene ID
1523
Gene Name
cut like homeobox 1
Gene Aliases
CASP, CDP, CDP/Cut, CDP1, COY1, CUTL1, CUX, Clox, Cux/CDP, GOLIM6, Nbla10317, p100, p110, p200, p75
UniGene
Hs.191482
Gene Chromosome Location
Chr.7: 101815904-102283958 on Build GRCh38
Transcripts
NM_001202543.1 NM_001202544.2 NM_001202545.2 NM_001202546.2 NM_001913.4 NM_181500.3 NM_181552.3 XM_005250150.2 XM_005250151.2 XM_005250154.3 XM_006715854.2 XM_006715855.1 XM_011515823.2 XM_011515825.1 XM_017011759.1 XM_017011760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5522464 c.89C>T p.S30F Chr.7: 101916173-101916173 on Build GRCh38
3083199 c.131A>G p.N44S Chr.7: 101916215-101916215 on Build GRCh38
3674786 c.170C>A p.P57Q Chr.7: 101916221-101916221 on Build GRCh38
1699089 c.169C>T p.P57S Chr.7: 101916220-101916220 on Build GRCh38
3662811 c.130C>T p.R44W Chr.7: 101916181-101916181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772151104 Chr.7: 101916276-101916276 on Build GRCh38
rs369222487 Chr.7: 101916244-101916244 on Build GRCh38
rs769308021 Chr.7: 101916138-101916138 on Build GRCh38
rs765983278 Chr.7: 101916243-101916243 on Build GRCh38
rs867880450 Chr.7: 101916133-101916133 on Build GRCh38
rs753731049 Chr.7: 101916123-101916123 on Build GRCh38
rs763823957 Chr.7: 101916216-101916216 on Build GRCh38
rs749792282 Chr.7: 101916180-101916180 on Build GRCh38
rs376836450 Chr.7: 101916132-101916132 on Build GRCh38
rs779416290 Chr.7: 101916270-101916270 on Build GRCh38
rs56091128 Chr.7: 101916233-101916233 on Build GRCh38
rs182847029 Chr.7: 101916277-101916277 on Build GRCh38
rs771086756 Chr.7: 101916072-101916072 on Build GRCh38
rs774765243 Chr.7: 101916078-101916078 on Build GRCh38
rs761704472 Chr.7: 101916221-101916221 on Build GRCh38
rs760414074 Chr.7: 101916215-101916215 on Build GRCh38
rs772195790 Chr.7: 101916193-101916193 on Build GRCh38
rs756778760 Chr.7: 101916124-101916124 on Build GRCh38
rs764022362 Chr.7: 101916111-101916111 on Build GRCh38
rs748511869 Chr.7: 101916213-101916213 on Build GRCh38
rs150487622 Chr.7: 101916147-101916147 on Build GRCh38
rs149191757 Chr.7: 101916218-101916218 on Build GRCh38
rs748548235 Chr.7: 101916240-101916240 on Build GRCh38
rs145317607 Chr.7: 101916189-101916189 on Build GRCh38
rs759647619 Chr.7: 101916080-101916080 on Build GRCh38
rs753599865 Chr.7: 101916258-101916258 on Build GRCh38
rs112658162 Chr.7: 101916097-101916097 on Build GRCh38
rs779860213 Chr.7: 101916243-101916244 on Build GRCh38
rs757861659 Chr.7: 101916237-101916237 on Build GRCh38
rs533781388 Chr.7: 101916235-101916235 on Build GRCh38
rs767411466 Chr.7: 101916084-101916084 on Build GRCh38
rs771176824 Chr.7: 101916181-101916181 on Build GRCh38
rs745939391 Chr.7: 101916272-101916272 on Build GRCh38
rs200309302 Chr.7: 101916182-101916182 on Build GRCh38
rs778253540 Chr.7: 101916249-101916249 on Build GRCh38
rs140709702 Chr.7: 101916117-101916117 on Build GRCh38
rs778368760 Chr.7: 101916128-101916128 on Build GRCh38
rs756596036 Chr.7: 101916246-101916246 on Build GRCh38
rs779702615 Chr.7: 101916146-101916146 on Build GRCh38
rs760459978 Chr.7: 101916107-101916107 on Build GRCh38
rs778447580 Chr.7: 101916159-101916159 on Build GRCh38
rs373330128 Chr.7: 101916250-101916250 on Build GRCh38
rs377325160 Chr.7: 101916241-101916241 on Build GRCh38

View Full Product Details